Skip to Content
Merck
All Photos(1)

Key Documents

1.06731

Sigma-Aldrich

Neocarmine MS "Fesago" color reagent

for the detection of textile fibres

Synonym(s):

Reagents for detection of textile fibres, Fesago

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$381.00

$381.00


Estimated to ship on27 April 2025


Request a Bulk Order

Select a Size

Change View
1 L
$381.00

About This Item

UNSPSC Code:
41116124
NACRES:
NA.21

$381.00


Estimated to ship on27 April 2025


Request a Bulk Order

Quality Level

form

liquid

pH

7.8 (20 °C in H2O)

density

1.01 g/cm3 at 20 °C

storage temp.

20-25°C

General description

for the detection of textile fibers

Application

For the detection of textile fibers

Analysis Note

Suitability for coloration of textiles:passes test

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service