Skip to Content
Merck
All Photos(1)

Documents

1.06663

Supelco

di-Sodium tartrate dihydrate

for analysis EMSURE®

Synonym(s):

di-Sodium tartrate dihydrate, di-Sodium tartrate dihydrate, Tartaric acid sodium salt dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4Na2O6 · 2H2O
CAS Number:
Molecular Weight:
230.08
MDL number:
UNSPSC Code:
12352107
EC Index Number:
212-773-3
NACRES:
NA.21

Quality Level

product line

EMSURE®

Assay

≥99.5% (perchloric acid titration)

form

powder

pH

8 (25 °C, 50 g/L in H2O)

solubility

290 g/L

density

1.82 g/cm3

bulk density

460 kg/m3

anion traces

chloride (Cl-): ≤0.0005%
phosphate (PO43-): ≤0.0005%
sulfate (SO42-): ≤0.002%

cation traces

Ca: ≤0.005%
Fe: ≤0.0005%
N: ≤0.002%
heavy metals (as Pb): ≤0.0005%

storage temp.

2-30°C

InChI

1S/C4H6O6.2Na.2H2O/c5-1(3(7)8)2(6)4(9)10;;;;/h1-2,5-6H,(H,7,8)(H,9,10);;;2*1H2/q;2*+1;;/p-2/t1-,2-;;;;/m1..../s1

InChI key

FGJLAJMGHXGFDE-DGFHWNFOSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Analysis Note

Assay (perchloric acid titration): ≥ 99.5 %
pH-value (5 %; water): 7.0 - 8.0
Chloride (Cl): ≤ 0.0005 %
Phosphate (PO₄): ≤ 0.0005 %
Sulfate (SO₄): ≤ 0.002 %
Total nitrogen (N): ≤ 0.002 %
Heavy metals (as Pb): ≤ 0.0005 %
Ca (Calcium): ≤ 0.005 %
Fe (Iron): ≤ 0.0005 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service