Skip to Content
Merck
All Photos(1)

Key Documents

1.06466

Supelco

Sodium hydroxide

monohydrate, 99.99 Suprapur®

Synonym(s):

Sodium hydroxide monohydrate, Soda caustic

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$206.00
500 G
$1,390.00

$206.00


Available to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
50 G
$206.00
500 G
$1,390.00

About This Item

Empirical Formula (Hill Notation):
HNaO · xH2O
CAS Number:
Molecular Weight:
40.00 (anhydrous basis)
MDL number:
UNSPSC Code:
12352305
NACRES:
NB.21

$206.00


Available to ship on31 March 2025Details


Request a Bulk Order

Product Name

Sodium hydroxide monohydrate, 99.99 Suprapur®

Quality Level

description

for inorganic trace analysis

Assay

99.99%

form

solid

pH

14 (20 °C, 50 g/L in H2O)

solubility

1100 g/L

storage temp.

2-30°C

SMILES string

[Na+].[O-H].O

InChI

1S/Na.2H2O/h;2*1H2/q+1;;/p-1

InChI key

JBJWASZNUJCEKT-UHFFFAOYSA-M

Related Categories

Analysis Note

Purity (metallic): ≥ 99.990 %
NaOH content: 67.0 - 69.0 %
Assay of sodium carbonate: ≤ 2.0 %
Chloride (Cl): ≤ 1 ppm
Phosphate (PO₄): ≤ 0.05 ppm
Silicate (as SiO₂): ≤ 5 ppm
Sulfate (SO₄): ≤ 10 ppm
Al (Aluminium): ≤ 0.05 ppm
As (Arsenic): ≤ 0.1 ppm
Ba (Barium): ≤ 10.0 ppm
Ca (Calcium): ≤ 0.50 ppm
Cd (Cadmium): ≤ 0.010 ppm
Ce (Cerium): ≤ 0.010 ppm
Co (Cobalt): ≤ 0.010 ppm
Cr (Chromium): ≤ 0.010 ppm
Cu (Copper): ≤ 0.010 ppm
Eu (Europium): ≤ 0.010 ppm
Fe (Iron): ≤ 0.05 ppm
Hg (Mercury): ≤ 0.01 ppm
K (Potassium): ≤ 2.0 ppm
La (Lanthanum): ≤ 0.010 ppm
Li (Lithium): ≤ 0.2 ppm
Mg (Magnesium): ≤ 0.10 ppm
Mn (Manganese): ≤ 0.010 ppm
Ni (Nickel): ≤ 0.010 ppm
Pb (Lead): ≤ 0.010 ppm
Sc (Scandium): ≤ 0.010 ppm
Sm (Samarium): ≤ 0.010 ppm
Sr (Strontium): ≤ 0.50 ppm
Tl (Thallium): ≤ 0.01 ppm
Y (Yttrium): ≤ 0.010 ppm
Yb (Ytterbium): ≤ 0.010 ppm
Zn (Zinc): ≤ 0.050 ppm

Legal Information

SUPRAPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service