Skip to Content
Merck
All Photos(1)

Key Documents

1063

Millipore

Acetylcoenzyme A, Trilithium Salt

Carrier of acyl groups in enzymatic reactions in fatty acid synthesis and oxidation and in biological acetylations.

Synonym(s):

Acetylcoenzyme A, Trilithium Salt

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.25
Assay:
≥83% (enzymatic)
form:
solid
solubility:
aqueous buffer, ph 4-6: 1 mg/mL
Pricing and availability is not currently available.

Quality Level

Assay

≥83% (enzymatic)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white

solubility

aqueous buffer, ph 4-6: 1 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

Carrier of acyl groups for enzymatic reactions involving fatty acid synthesis and oxidation and in biological acetylations.
Carrier of acyl groups in enzymatic reactions in fatty acid synthesis and oxidation and in biological acetylations.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Aqueous stock solutions are stable for up to 1-2 weeks at -20°C.

Other Notes

Gorst, C.M., and Ragsdale, S.W. 1991. J. Biol. Chem.266, 20687.
Power, R., et al. 1990. Arch. Biochem. Biophys.279, 370.
Mannens, G., et al. 1988. Biochim. Biophys. Acta959, 214.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service