Skip to Content
Merck
All Photos(3)

Documents

1.06059

Supelco

Methyl benzoate

EMPLURA®

Synonym(s):

Methyl benzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8O2
CAS Number:
Molecular Weight:
136.15
MDL number:
UNSPSC Code:
12191501
EC Index Number:
202-259-7
NACRES:
NA.05

vapor pressure

0.36 hPa ( 20 °C)

Quality Level

product line

EMPLURA®

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

510 °C

potency

1177 mg/kg LD50, oral (Rat)
2170 mg/kg LD50, skin (Rabbit)

expl. lim.

8.6-20 % (v/v)

impurities

≤0.05% Sulfated ash
≤0.1% Water
≤0.2% Free acid (as benzoic acid)

bp

199 °C/1013 hPa

mp

-12 °C

transition temp

flash point 82 °C

density

1.09 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C8H8O2/c1-10-8(9)7-5-3-2-4-6-7/h2-6H,1H3

InChI key

QPJVMBTYPHYUOC-UHFFFAOYSA-N

Related Categories

General description

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.

Analysis Note

Purity (GC): ≥ 99.0 %
Identity (IR): conforms
Free acid (as benzoic acid): ≤ 0.2 %
Density (d 20 °C/ 4 °C): 1.086 - 1.088
Sulfated ash: ≤ 0.05 %
Water: ≤ 0.1 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

170.6 °F - Pensky-Martens closed cup

Flash Point(C)

77 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service