Skip to Content
Merck
All Photos(1)

Documents

1.05957

Supelco

Manganese(IV) oxide

powder EMPLURA®

Synonym(s):

Manganese(IV) oxide, Manganese dioxide, Pyrolusite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
MnO2
CAS Number:
Molecular Weight:
86.94
MDL number:
UNSPSC Code:
12352303
EC Index Number:
215-202-6
NACRES:
NA.21

Quality Level

product line

EMPLURA®

Assay

≥89.0% (manganometric)

form

powder

potency

>3478 mg/kg LD50, oral (Rat)

impurities

≤5% Substances insoluble in hydrochloric acid

loss

≤0.3% loss on drying, 105°C

solubility

insoluble <0.001 g/L

density

5.21 g/cm3 at 21 °C

bulk density

600‑800 kg/m3

anion traces

sulfate (SO42-): ≤0.1%

cation traces

Ba: ≤1.0%
Ca: ≤1.0%
Fe: ≤1.5%

storage temp.

2-30°C

InChI

1S/Mn.2O

InChI key

NUJOXMJBOLGQSY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application



  • The chemically synthesized ageladine A-derivative LysoGlow84 stains lysosomes in viable mammalian brain cells and specific structures in the marine flatworm Macrostomum lignano.: This research highlights the use of a manganese-based synthetic derivative, LysoGlow84, which effectively stains lysosomes in mammalian brain cells and certain marine flatworms. The study underscores its significance in cellular biology and molecular imaging, providing a tool for detailed lysosomal studies in diverse biological systems (Mordhorst et al., 2015).


Analysis Note

Assay (manganometric): ≥ 89.0 %
Substances insoluble in hydrochloric acid: ≤ 5.0 %
Sulfate (SO₄): ≤ 0.1 %
Ba (Barium): ≤ 1.0 %
Ca (Calcium): ≤ 1.0 %
Fe (Iron): ≤ 1.5 %
Loss on Drying (105°C): ≤ 0.3 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - STOT RE 2 Inhalation

Target Organs

Brain

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service