Skip to Content
Merck
All Photos(1)

Key Documents

1.05153

Supelco

Potassium sulfate

for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Synonym(s):

Potassium sulfate, Potassium pyrosulfate

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$80.20
1 KG
$124.00
5 KG
$432.00
25 KG
$2,470.00

$80.20


Available to ship on07 April 2025Details


Request a Bulk Order

Select a Size

Change View
500 G
$80.20
1 KG
$124.00
5 KG
$432.00
25 KG
$2,470.00

About This Item

Empirical Formula (Hill Notation):
K2O4S
CAS Number:
Molecular Weight:
174.26
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-915-5
NACRES:
NA.21
grade:
ACS reagent
form:
solid

$80.20


Available to ship on07 April 2025Details


Request a Bulk Order

grade

ACS reagent

Quality Level

Agency

reag. ISO
reag. Ph. Eur.

product line

EMSURE®

Assay

≥99.0% (alkalimetric)

form

solid

potency

6600 mg/kg LD50, oral (Rat)

impurities

≤0.01% Insoluble matter

pH

5.5-8.5 (20 °C, 50 g/L in H2O)

bp

1689 °C/1013 hPa

mp

1069 °C

density

2.66 g/cm3 at 20 °C

bulk density

800 kg/m3

anion traces

chloride (Cl-): ≤0.0005%

cation traces

As: ≤0.0002%
Ca: ≤0.005%
Fe: ≤0.0005%
Mg: ≤0.002%
N: ≤0.0005%
Na: ≤0.02%
heavy metals (as Pb): ≤0.0005%

storage temp.

2-30°C

SMILES string

[K+].[K+].[S](=O)(=O)([O-])[O-]

InChI

1S/2K.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

OTYBMLCTZGSZBG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

  • Fertilizer application alters cadmium and selenium bioavailability in soil-rice system with high geological background levels: This study explores how different fertilizer applications, including potassium sulfate, affect the bioavailability of cadmium and selenium in rice cultivated in areas with naturally high levels of these elements (Zhou et al., 2024).
  • Exogenous application of nano-silicon, potassium sulfate, or proline enhances physiological parameters, antioxidant enzyme activities, and agronomic traits of diverse rice genotypes under water deficit conditions: Research indicates that potassium sulfate, along with nano-silicon and proline, can improve water stress resilience in rice by enhancing physiological traits and antioxidant activities (Abd-El-Aty et al., 2024).
  • Size-Independent Nucleation and Growth Model of Potassium Sulfate from Supersaturated Solution Produced by Stirred Crystallization: This paper presents a model of potassium sulfate crystallization that is independent of crystal size, providing insights into the controlled crystallization processes used in industry (Zheng, 2023).
  • Prediction models of macro-nutrient content in plant organs of Cucumis melo in response to soil elements using support vector regression: This study includes potassium sulfate among the soil elements analyzed for their impact on nutrient uptake in melon, with models predicting how different soil treatments affect plant growth (Keshtehgar et al., 2023).
  • [Effects of chemical fertilizer reduction and organic fertilizer substitution on yield and quality of Saposhnikovia divaricata]: This research evaluates the effects of reducing chemical fertilizers, including potassium sulfate, and substituting with organic options, highlighting changes in plant yield and quality (Li et al., 2023).

Linkage

Replaces: PX1595; PX1595-1; PX1595-5; PX1595-6; PX1595-20

Analysis Note

Assay (alkalimetric): ≥ 99.0 %
Insoluble matter: ≤ 0.01 %
pH-value (5 %; water, 25 °C): 5.5 - 7.5
Chloride (Cl): ≤ 0.0005 %
Total nitrogen (N): ≤ 0.0005 %
Heavy metals (as Pb): ≤ 0.0005 %
As (Arsenic): ≤ 0.0002 %
Ca (Calcium): ≤ 0.005 %
Fe (Iron): ≤ 0.0005 %
Mg (Magnesium): ≤ 0.002 %
Na (Sodium): ≤ 0.02 %
Corresponds to ACS,ISO,Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service