Skip to Content
Merck
All Photos(1)

Key Documents

1.05073

Supelco

di-Potassium oxalate monohydrate

for analysis EMSURE® ACS

Synonym(s):

di-Potassium oxalate monohydrate, Oxalic acid dipotassium salt monohydrate

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$177.00
1 KG
$451.00

$177.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
250 G
$177.00
1 KG
$451.00

About This Item

Empirical Formula (Hill Notation):
C2K2O4 · xH2O
CAS Number:
Molecular Weight:
166.22 (anhydrous basis)
MDL number:
UNSPSC Code:
12352107
EC Index Number:
209-506-8
NACRES:
NA.21
grade:
ACS reagent
form:
solid
solubility:
392 g/L (anhydrous substance)

$177.00


Estimated to ship on02 June 2025


Request a Bulk Order

grade

ACS reagent

Quality Level

vapor pressure

<0.1 hPa ( 20 °C)

product line

EMSURE®

Assay

99.5-101.0% (oxidimetric)

form

solid

potency

660 mg/kg LD50, oral (Rat)

impurities

≤0.01% Insoluble matter

pH

7.0-8.5 (20 °C, 50 g/L in H2O)

mp

>138 °C (decomposition)

solubility

392 g/L (anhydrous substance)

density

2.20 g/cm3 at 20.4 °C

bulk density

700‑1100 kg/m3

anion traces

chloride (Cl-): ≤0.001%
sulfate (SO42-): ≤0.01%

cation traces

Fe: ≤0.0005%
NH4+: ≤0.002%
Na: ≤0.02%
heavy metals (as Pb): ≤0.001%

storage temp.

2-30°C

SMILES string

[K+].[K+].[O-]C(=O)C(=O)[O-].O

InChI

1S/C2H2O4.2K.H2O/c3-1(4)2(5)6;;;/h(H,3,4)(H,5,6);;;1H2/q;2*+1;/p-2

InChI key

QCPTVXCMROGZOL-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Analysis Note

Assay (oxidimetric): 99.5 - 101.0 %
Insoluble matter: ≤ 0.01 %
Neutrality: passes test
Chloride (Cl): ≤ 0.001 %
Sulfate (SO₄): ≤ 0.01 %
Heavy metals (as Pb): ≤ 0.001 %
NH₄ (Ammonium): ≤ 0.002 %
Fe (Iron): ≤ 0.0005 %
Na (Sodium): ≤ 0.02 %
Substances darkened by hot sulfric acid: passes test
Corresponds to ACS

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service