Skip to Content
Merck
All Photos(1)

Key Documents

1.04410

Supelco

Mercury(II) acetate

for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

Mercury(II) acetate

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$144.00
250 G
$299.00

$144.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
50 G
$144.00
250 G
$299.00

About This Item

Linear Formula:
Hg(CH3COO)2
CAS Number:
Molecular Weight:
318.68
MDL number:
UNSPSC Code:
12352103
EC Index Number:
216-491-1
NACRES:
NA.21

$144.00


Estimated to ship on28 May 2025


Request a Bulk Order

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

product line

EMSURE®

Assay

≥99.0% (complexometric)

form

solid

potency

40.9 mg/kg LD50, oral (Rat)

impurities

≤0.01% Substances insoluble in dilute acetic acid

reduction residue

≤0.02%

mp

178-180 °C (decomposition)

density

3.27 g/cm3 at 20 °C

bulk density

1000 kg/m3

anion traces

chloride (Cl-): ≤0.005%
nitrate (NO3-): ≤0.005%
sulfate (SO42-): ≤0.005%

cation traces

Fe: ≤0.001%
Hg+: ≤0.3%
other heavy metals (as Pb): ≤0.002%

storage temp.

2-30°C

SMILES string

[Hg+2].[O-]C(=O)C.[O-]C(=O)C

InChI

1S/2C2H4O2.Hg/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+2/p-2

InChI key

BRMYZIKAHFEUFJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Stable -ESiMe3 Complexes of Cu(I) and Ag(I) (E=S, Se) with NHCs: Synthons in Ternary Nanocluster Assembly.: This research discusses the synthesis and characterization of stable Cu(I) and Ag(I) complexes, possibly applicable in nanocluster assembly, which includes Mercury(II) acetate as a catalyst for synthesis processes (Azizpoor Fard et al., 2016).

  • Zinc(II), cadmium(II), mercury(II), and ethylmercury(II) complexes of phosphinothiol ligands.: This article explores the coordination chemistry of mercury(II) with phosphinothiol ligands, offering insights into the complexation behavior that can influence analytical methodologies in detecting mercury species (Fernández et al., 2008).

  • Microscale preparation of even- and odd-numbered N-acetylheparosan oligosaccharides.: This study includes the use of Mercury(II) acetate in the enzymatic synthesis processes of oligosaccharides, highlighting its role in the preparation of complex biochemical structures (Minamisawa et al., 2006).

Analysis Note

Assay (complexometric): ≥ 99.0 %
Substances insoluble in dilute acetic acid: ≤ 0.01 %
Chloride (Cl): ≤ 0.005 %
Nitrate (NO₃): ≤ 0.005 %
Sulfate (SO₄): ≤ 0.005 %
other heavy metals (as Pb): ≤ 0.002 %
Fe (Iron): ≤ 0.001 %
Mercury (I) (as Hg): ≤ 0.3 %
Residue after reduction: ≤ 0.02 %
Correspons to ACS,Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service