Skip to Content
Merck
All Photos(1)

Documents

1.04324

Millipore

NutriSelect® prime Plate Count Agar halal acc. ISO 4833, ISO 17410 and FDA-BAM

Suitable for use in Halal and Kosher certified processes, NutriSelect® Prime

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

Agency

AOAC® 940.36
APHA
GB 4789.2
GB 4789.43
ISO 17410
ISO 4833-1
ISO 4833-2

Quality Level

reg. compliance

FDA BAM Medium M124

form

powder

manufacturer/tradename

NutriSelect® Prime

technique(s)

microbiological culture: suitable

pH

7.0 (30.0 °C)

application(s)

food and beverages

storage temp.

15-25°C

suitability

aerobic bacteria

Related Categories

General description

NutriSelect® prime Plate Count Agar halal is a powdered culture medium without any inhibitors or indicators, and it is relatively rich in its nutrients. The enzymatic digest of casein (tryptone) with a high level of free amino acids is a nitrogen source. Yeast extract primarily supplies the B-complex vitamins. Glucose is an energy source for the growth of bacteria, while agar is the solidifying agent.

Application

NutriSelect® prime Plate Count Agar halal is suitable for the determination of the total microbial content from milk, dairy products, food and animal feed, water, beverages, and other materials.

Legal Information

AOAC is a registered trademark of AOAC International
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Halal-certified culture media prevent non-halal ingredient contamination, enabling microbiological QC testing without losing certification.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service