Skip to Content
Merck

1.04166

SAFC

Urea

granulated, EMPROVE® EXPERT, Ph. Eur., BP, ChP, JP, USP, ACS

Pharma Manufacturing

Synonym(s):

Carbamide, Carbonyldiamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CONH2
CAS Number:
Molecular Weight:
60.06
Beilstein:
635724
MDL number:
UNSPSC Code:
12352111
Pricing and availability is not currently available. Contact Technical Service

grade

ACS reagent

Quality Level

Agency

BP
ChP
JP
Ph. Eur.
USP

product line

EMPROVE® EXPERT

form

granular

pH

9

mp

132-135 °C (lit.)

density

1.335 g/mL at 25 °C (lit.)

application(s)

liquid formulation
parenterals
pharma/biopharma processes
pharmaceutical

shipped in

ambient

SMILES string

NC(N)=O

InChI

1S/CH4N2O/c2-1(3)4/h(H4,2,3,4)

InChI key

XSQUKJJJFZCRTK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Standard bulk materials often show a tendency for caking, clumping and dust formation which results in laborious weighing processes and manual de-caking processes with a negative impact on manufacturing efficiency. Due to its granulated form, granulated urea is free-flowing and easy to handle. Its greatly reduced caking behavior facilitates processability, speeds up manufacturing processes and increases operator safety.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Urea is typically used for the denaturation of proteins and as a mild solubilization agent for insoluble or denatured proteins. It is useful for renaturing proteins from samples already denatured with 6 M guanidine chloride such as inclusion bodies. Urea may be used with guanidine hydrochloride and dithiothreitrol (DTT) in the refolding of denatured proteins into their native or active form.
Due to its low microbial and endotoxin limits, Urea granulated Emprove® Expert is especially suitable for high-risk applications.
Used for the denaturation of proteins and as a mild solubilization agent for insoluble or denatured proteins. Useful for renaturing proteins from samples already denatured with 6 M guanidine chloride such as inclusion bodies. May be used with guanidine hydrochloride and dithiothreitrol (DTT) in the refolding of denatured proteins into their native or active form.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Application

Product No.
Description
Pricing

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Handling bulk powders at a large scale in the pharmaceutical, biopharmaceutical, and bioprocessing setting is challenging. Compared to powders, granulated materials experience far less caking, have better flowability, and are easier to handle, even after long-term storage.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service