Skip to Content
Merck
All Photos(1)

Key Documents

1.04018

Supelco

Glass beads 6 mm

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$130.00

$130.00


Available to ship on03 April 2025Details


Request a Bulk Order

Select a Size

Change View
500 G
$130.00

About This Item

UNSPSC Code:
41122409
NACRES:
NB.51

$130.00


Available to ship on03 April 2025Details


Request a Bulk Order

storage temp.

no temp limit

Quality Level

Application


  • Effect of an innovative sorbent material coupled to continuous flow process in the protein and oxidative stability of white wines.: This research demonstrates the use of glass beads as part of an innovative sorbent system to improve the stability and quality of white wines, highlighting its practical applications in food chemistry (Ricci et al., 2024).

  • Thin-section- and matrix-free mass spectrometry imaging: Reproducible sample transfer using novel platinum-coated porous plate formed of glass beads.: This study introduces a new application of glass beads in mass spectrometry, creating a platform that enhances the reproducibility and efficiency of sample analysis, crucial for precise chemical imaging (Ikeda et al., 2024).

  • Raman Spectroscopy of Glass Beads in Ammonium Nitrate Solution and Compensation of Signal Losses.: This paper explores the use of glass beads to refine Raman spectroscopy techniques, specifically in identifying and compensating for signal losses in complex solutions, which is vital for accurate analytical measurements (Spoor et al., 2024).

  • Force on a sphere suspended in flowing granulate.: This research utilizes glass beads to study the dynamics of granular flows around suspended objects, providing insights into the physical interactions and forces in granular materials, which can be applied to various industrial and research settings (Wang et al., 2023).

  • Micro computed tomography images of capillary actions in rough and irregular granular materials.: This study uses glass beads to simulate and study capillary actions in granular materials through micro computed tomography, enhancing understanding of fluid dynamics in porous media, relevant in both environmental science and engineering (Nadimi et al., 2024).


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service