Skip to Content
Merck
All Photos(1)

Documents

1.03854

Millipore

GranuCult® prime Thiosulfate Citrate Bile Sucrose (TCBS) Agar

acc. FDA-BAM, ISO 21872

Synonym(s):

TCBS Agar, Thiosulfate Citrate Bile Salts Sucrose Agar, Vibrio Selective Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

Agency

ISO 21872

Quality Level

form

powder

technique(s)

microbiological culture: suitable

pH

8.6±0.2 (25 °C)

application(s)

food and beverages
pharmaceutical

storage temp.

15-25°C

suitability

selective and differential for Vibrio spp.

General description

GranuCult® prime Thiosulfate Citrate Bile Sucrose (TCBS) Agar is a complex medium designed for the isolation and differentiation of Vibrio spp. and other enteropathogenic microorganisms from products intended for human consumption, animal feed, environmental samples in the production area, food production, food handling, and other materials.
The synergistic effect of high concentration of thiosulfate, citrate and strong alkalinity inhibits the growth of Enterobacteriaceae spp. The medium is highly selective as the bile salts present in the medium suppresses the growth of Enterococci spp.
The differentiation is achieved based on sucrose fermentation, pH indicators and H2S production from thiosulfate. Vibrio spp. do not produce H2S but depending on the species, may ferment sucrose. Peptone and yeast extract in the media allow good growth and agar is the solidifying agent.

Application

GranuCult® prime Thiosulfate Citrate Bile Sucrose (TCBS) Agar is used to isolate and differentiate Vibrio spp. and other enteropathogenic microorganisms from products intended for human consumption, animal feed, environmental samples in the production area, food production, food handling, and other materials.

Analysis Note

  • The dehydrated medium is a granulate with green color.
  • The prepared medium is clear to slightly opalescent and blue-green.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service