Skip to Content
Merck
All Photos(3)

Key Documents

1.03771

Supelco

Ethanol for analysis

completely denatured with 1% Ethyl methyl ketone, 1% Isopropyl alcohol, 1g/100l Denatonium benzoate EMSURE®, for extraction, denatured

Synonym(s):

Analytical-Grade Ethanol

Sign Into View Organizational & Contract Pricing

Select a Size

2.5 L
$81.90

$81.90


Available to ship on28 March 2025Details


Request a Bulk Order

Select a Size

Change View
2.5 L
$81.90

About This Item

UNSPSC Code:
12191502
NACRES:
NB.21
grade:
for extraction
application(s):
environmental
food and beverages
industrial qc
pharmaceutical
sample preparation

$81.90


Available to ship on28 March 2025Details


Request a Bulk Order

grade

for extraction

description

for analytical purposes

product line

EMSURE®

Assay

≥99.5% (GC)

form

liquid

contains

0.01 g/L Denatonium benzoate as denaturant
1% Ethyl methyl ketone as denaturant
1% Isopropyl alcohol as denaturant

dilution

(for analytical testing)

impurities

≤0.000002% Co (Cobalt)
≤0.000002% Cr (Chromium)
≤0.000002% Cu (Copper)
≤0.000002% Mn (Manganese)
≤0.000002% Ni (Nickel)
≤0.000005% Cd (Cadmium)
≤0.00001% Ba (Barium)
≤0.00001% Fe (Iron)
≤0.00001% Mg (Magnesium)
≤0.00001% Pb (Lead)
≤0.00001% Sn (Tin)
≤0.00001% Zn (Zinc)
≤0.00005% Al (Aluminium)
≤0.00005% B (Boron)
≤0.00005% Ca (Calcium)
≤0.0001% K (Potassium)
≤0.003% Free acid (as acetic acid)
≤0.1% Water
0.8-1.3% Ethylmethylketone (GC)
0.8-1.3% Isopropyl alcohol (GC)

color

APHA: ≤10

transition temp

flash point 12 °C

density

0.79 g/cm3 at 20 °C

application(s)

environmental
food and beverages
industrial qc
pharmaceutical
sample preparation

format

neat

storage temp.

2-30°C

General description

completely denatured with 1% Ethyl methyl ketone, 1% Isopropyl alcohol, 1g/100l Denatonium benzoate EMSURE®

Application


  • Chemical constituents from the Saposhnikovia divaricata and their antiproliferative activity.: This study utilized ethanol for analysis in the extraction and identification of chemical constituents from Saposhnikovia divaricata, which demonstrated significant antiproliferative activity against cancer cells (Sun et al., 2024).

  • In Vitro Bioactivities and Characterization of Mycelial Extracts from Different Strains of Phellinus igniarius (Agaricomycetes).: The research employed ethanol for analysis to extract bioactive compounds from various strains of Phellinus igniarius, showcasing their potential medicinal properties (Dong et al., 2024).

  • Antitussive, Expectorant and Antipyretic Effect of the Ethanolic Extract of the Leaves of Momordica charantia L.: This study used ethanol for analysis to prepare extracts from Momordica charantia leaves, which exhibited significant antitussive, expectorant, and antipyretic effects (Ribeiro et al., 2024).

  • Optimization of guar gum-based anti-browning coating for prolonging the shelf life of cut potatoes.: Ethanol for analysis was utilized in optimizing a guar gum-based coating, effectively extending the shelf life of cut potatoes by preventing browning (Mondal et al., 2024).

Linkage

Discover EMSURE - to be more than sure!

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F - Ethanol

Flash Point(C)

12 °C - Ethanol


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service