Skip to Content
Merck
All Photos(1)

Key Documents

1.03062

Supelco

Dimethylglyoxime

GR for analysis (reagent for nickel) ACS,Reag. Ph Eur

Synonym(s):

Dimethylglyoxime, Diacetyldioxime, 2,3-Butanedionedioxime disodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$360.00

$360.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
100 G
$360.00

About This Item

Linear Formula:
(CH3CNOH)2
CAS Number:
Molecular Weight:
116.12
MDL number:
UNSPSC Code:
12352126
EC Index Number:
202-420-1
NACRES:
NA.05
grade:
ACS reagent

$360.00


In StockDetails


Request a Bulk Order

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

Assay

≥99% (gravimetric)

form

solid

dilution

(for analytical testing)

mp

239-243 °C (decomposition)

bulk density

620 kg/m3

storage temp.

2-30°C

SMILES string

N(O)\C(=C(/N=O)\C)\C

InChI

1S/C4H8N2O2/c1-3(5-7)4(2)6-8/h5,7H,1-2H3/b4-3-

InChI key

OFZZNQXYRRSXOI-ARJAWSKDSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (gravimetrically): ≥ 99
Identity (IR-spectrum): passes test
Matter insoluble in ethanol: ≤ 0.02 %
Melting range: 239 - 243 °C
Sulfated ash: ≤ 0.05 %
Suitability as reagent (at Nickel): passes test

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service