Skip to Content
Merck
All Photos(1)

Key Documents

1.02733

Supelco

Copper(II) chloride dihydrate

for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

Copper(II) chloride dihydrate, Copper dichloride dihydrate

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$122.00
1 KG
$305.00

$122.00


Available to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
250 G
$122.00
1 KG
$305.00

About This Item

Empirical Formula (Hill Notation):
Cl2Cu · 2H2O
CAS Number:
Molecular Weight:
170.48
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-210-2
NACRES:
NA.21
grade:
ACS reagent
form:
solid
solubility:
757 g/L

$122.00


Available to ship on31 March 2025Details


Request a Bulk Order

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

product line

EMSURE®

Assay

≥99.0% (iodometric)

form

solid

potency

584 mg/kg LD50, oral (Rat)

impurities

≤0.01% Insoluble matter

pH

3.0-3.8 (20 °C, 50 g/L in H2O)

mp

598 °C

solubility

757 g/L

density

2.53 g/cm3 at 20 °C

bulk density

1070 kg/m3

anion traces

nitrate (NO3-): ≤0.015%
sulfate (SO42-): ≤0.005%

cation traces

As: ≤0.0001%
Ca: ≤0.005%
Fe: ≤0.003%
K: ≤0.01%
N: ≤0.003%
Na: ≤0.02%
Ni: ≤0.001%
Pb: ≤0.004%

storage temp.

2-30°C

SMILES string

[Cu](Cl)Cl.O.O

InChI

1S/2ClH.Cu.2H2O/h2*1H;;2*1H2/q;;+2;;/p-2

InChI key

MPTQRFCYZCXJFQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Chitosan-based film supported copper nanoparticles: A potential and reusable catalyst for the reduction of aromatic nitro compounds.: This study investigates the application of copper(II) chloride dihydrate in the development of chitosan-based films embedded with copper nanoparticles, which serve as an effective and reusable catalyst for the reduction of aromatic nitro compounds. The process highlights the potential of copper(II) chloride dihydrate in catalytic applications within analytical chemistry, providing a sustainable approach to chemical reactions (de Souza JF et al., 2017).

Linkage

Replaces: CX2150; CX2150-1; CX2150-5

Analysis Note

Assay (iodometric): ≥ 99.0 %
Insoluble matter: ≤ 0.01 %
pH-value (5 %; water): 3.0 - 3.8
Sulfate (SO₄): ≤ 0.005 %
Nitrate (NO₃): ≤ 0.015 %
Total nitrogen (N): ≤ 0.003 %
As (Arsenic): ≤ 0.0001 %
Ca (Calcium): ≤ 0.005 %
Fe (Iron): ≤ 0.003 %
K (Potassium): ≤ 0.01 %
Na (Sodium): ≤ 0.02 %
Ni (Nickel): ≤ 0.001 %
Pb (Lead): ≤ 0.004 %
Corresponds to ACS, Reag. Ph. Eur.

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service