Skip to Content
Merck
All Photos(1)

Key Documents

1.02473

Sigma-Aldrich

ISOSLIDE Methenamine Control Slides

control slides with reference tissue for the detection of argent-affine structures in histological tissue

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116124
Pricing and availability is not currently available.

Quality Level

IVD

for in vitro diagnostic use

application(s)

clinical testing
diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

General description

The ISOSLIDE Methenamine Control Slides - with reference tissue for the detection of argent-affine structures in histological tissue, are prepared using sections of suitable animal material, that clearly visualize argent-affin structures in enterochromaffin cells and basal membranes. Each package of ISOSLIDE control slides contains 25 slides. The tissue is applied onto the slide next to the white printed labelling field. One slide is already stained with the reference method (Methenamine silver plating kit acc. to Gomori, Product number 1.00820) and is used for comparison. The 24 unstained slides are stained according to the corresponding protocol or with the laboratory′s own protocol. After the staining of the control slides, the staining result is to be compared with the laboratory material and the pre-stained control slide. The ISOSLIDE control slides are not quality-controlled for their application with molecular pathology methods and IHC.
This product is registered as IVD and CE marked. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Application

Control slides with reference tissue for the detection of argentaffin structures in histological tissue

Legal Information

ISOSLIDE is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service