Skip to Content
Merck
All Photos(1)

Key Documents

1.02331

Millipore

Cellulose microcrystalline

for column chromatography

Synonym(s):

Cellulose microcrystalline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H10O5)n
CAS Number:
MDL number:
UNSPSC Code:
41115711
EC Index Number:
232-674-9
Pricing and availability is not currently available. Contact Technical Service

material

cellulose matrix
plastic support

Quality Level

form

microcrystalline

autoignition temp.

232 °C

potency

>5000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

feature

binder Organic Polymer
fluorescent indicator: no

packaging

pkg of 500 g

technique(s)

thin layer chromatography (TLC): suitable

particle size

<20 μm

pore size

60 Å medium pore diameter

pH

5-7.5 (20 °C, 100 g/L in H2O, slurry)

transition temp

flash point 260 °C

density

1.5 g/cm3 at 20 °C

bulk density

70‑400 kg/m3

storage temp.

no temp limit

SMILES string

O1[C@H](C(C(C(C1CO)O)O)O)O[C@@H]2C(OC(C(C2O)O)O)CO

InChI

1S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20H,1-2H2/t3?,4?,5?,6?,7?,8?,9?,10-,11?,12+/m1/s1

InChI key

GUBGYTABKSRVRQ-WFVLMXAXSA-N

Analysis Note

Sieve analysis (<20 µm): ≤20%
Sieve analysis (>160 µm): ≤2%
Sieve analysis (20-160 µm): ≥80%

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service