Skip to Content
Merck
All Photos(1)

Key Documents

1.02126

Millipore

α-Cyclodextrine

for biochemistry

Synonym(s):

α-Cyclodextrine, Schardinger α-dextrin, Cyclohexaamylose, Cyclomaltohexaose

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$353.00

$353.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
50 G
$353.00

About This Item

Empirical Formula (Hill Notation):
C36H60O30
CAS Number:
Molecular Weight:
972.84
MDL number:
UNSPSC Code:
12352201
EC Index Number:
233-007-4
NACRES:
NA.79

$353.00


Estimated to ship on26 May 2025


Request a Bulk Order

Quality Level

Assay

≥98.0% (HPLC)

form

solid

potency

>10000 mg/kg LD50, oral (Rat)

mp

278 °C

bulk density

500 kg/m3

storage temp.

15-25°C

SMILES string

O1C2OC(C(OC3OC(C(OC4OC(C(OC5OC(C(OC6OC(C(OC7OC(C1C(C7O)O)CO)C(C6O)O)CO)C(C5O)O)CO)C(C4O)O)CO)C(C3O)O)CO)C(C2O)O)CO

InChI

1S/C36H60O30/c37-1-7-25-13(43)19(49)31(55-7)62-26-8(2-38)57-33(21(51)15(26)45)64-28-10(4-40)59-35(23(53)17(28)47)66-30-12(6-42)60-36(24(54)18(30)48)65-29-11(5-41)58-34(22(52)16(29)46)63-27-9(3-39)56-32(61-25)20(50)14(27)44/h7-54H,1-6H2

InChI key

HFHDHCJBZVLPGP-UHFFFAOYSA-N

Analysis Note

Purity (HPLC): ≥ 98.0 %
Identity (IR-spectrum): conforms
Water (according to Karl Fischer): ≤ 11.0 %
Spec. rotation [α²0/D (1 %; water; calc. on anhydrous substance): +148.0 - +153.0 °

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service