Skip to Content
Merck
All Photos(2)

Documents

1.01786

Supelco

Petroleum benzine boiling range 30-50°C

for analysis EMSURE®

Synonym(s):

Petroleum ether, Petroleum spirit, Petroleum ether

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12191502
NACRES:
NA.21

Quality Level

product line

EMSURE®

form

liquid

autoignition temp.

285 °C

expl. lim.

1.2-7.5 % (v/v)

impurities

≤0.000002% B (Boron)
≤0.00005% Al (Aluminium)
≤0.0002 meq/g Acidity
≤0.005% Sulfur compounds (as S)
≤0.01% Aromatics (as benzene)
≤0.01% Water

evapn. residue

≤0.003%

color

APHA: ≤10

bp

30-50 °C/1013 hPa

iodine value

≤0.3

transition temp

flash point <-30 °C

density

0.65 g/cm3 at 20 °C

cation traces

Ba: ≤0.00001%
Ca: ≤0.00005%
Cd: ≤0.000005%
Co: ≤0.000002%
Cr: ≤0.000002%
Cu: ≤0.000002%
Fe: ≤0.00001%
Mg: ≤0.00001%
Mn: ≤0.000002%
Ni: ≤0.000002%
Pb: ≤0.00001%
Sn: ≤0.00001%
Zn: ≤0.00001%

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

General description

EMSURE grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE provides worldwide best and most extensive product specifications. We declare our EMSURE range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Analysis Note

Acidity: ≤ 0.0002 meq/g
Colour: ≤ 10 Hazen
Boiling range: 30 - 50 °C
Aromatics (as benzene): ≤ 0.01 %
Sulfur compounds (as S): ≤ 0.005 %
Iodine number: ≤ 0.3
Evaporation residue: ≤ 0.003 %
Water: ≤ 0.01 %
Al (Aluminium): ≤ 0.00005 %
B (Boron): ≤ 0.000002 %
Ba (Barium): ≤ 0.00001 %
Ca (Calcium): ≤ 0.00005 %
Cd (Cadmium): ≤ 0.000005 %
Co (Cobalt): ≤ 0.000002 %
Cr (Chromium): ≤ 0.000002 %
Cu (Copper): ≤ 0.000002 %
Fe (Iron): ≤ 0.00001 %
Mg (Magnesium): ≤ 0.00001 %
Mn (Manganese): ≤ 0.000002 %
Ni (Nickel): ≤ 0.000002 %
Pb (Lead): ≤ 0.00001 %
Sn (Tin): ≤ 0.00001 %
Zn (Zinc): ≤ 0.00001 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Carc. 1B - Flam. Liq. 1 - Muta. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-22.0 °F

Flash Point(C)

< -30 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service