Skip to Content
Merck
All Photos(1)

Key Documents

1.01632

Supelco

Monochloramine Test, photometric

photometric, 0.050-10.00 mg/L (Cl2), 0.036-7.26 mg/L (NH2Cl), 0.010-1.98 mg/L (NH2Cl-N), Spectroquant®

Sign Into View Organizational & Contract Pricing

Select a Size

150 TESTS
$504.00

$504.00


Available to ship on31 March 2025Details


Request a Bulk OrderRequest more information

Select a Size

Change View
150 TESTS
$504.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

$504.00


Available to ship on31 March 2025Details


Request a Bulk OrderRequest more information

Product Name

Monochloramine Test, photometric, 0.050-10.00 mg/L (Cl2), 0.036-7.26 mg/L (NH2Cl), 0.010-1.98 mg/L (NH2Cl-N), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 150 tests

specific analyte(s)

monochloramine

measuring range

0.010-1.98 mg/L (NH2Cl-N)
0.036-7.26 mg/L (NH2Cl)
0.050-10.00 mg/L (Cl2)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Indophenol blue)

storage temp.

15-25°C

General description

Method: photometric 0.050 - 10.00 mg/l Cl₂ 0.036 - 7.26 mg/l NH₂Cl 0.010 - 1.98 mg/l NH₂Cl-N Spectroquant®

This Spectroquant® Monachloramine Reagent Test allows the accurate quantification of the monochloramine content in water samples and disinfectant solutions.

Method applied: In the presence of a catalyst, monochloramine (NH2Cl) reacts with thymol to form a blue indophenol derivative which is determined photometrically.

The Spectroquant® Reagent Test Kits contain highly stable, ready-to-use reagent mixtures to perform the analysis according to the procedure described in the accompanying instruction leaflet.

All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service