Skip to Content
Merck
All Photos(2)

Key Documents

1.01512

Supelco

Silver nitrate

for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Synonym(s):

Nitric acid silver(I) salt

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$301.00
100 G
$695.00
250 G
$1,670.00
1 KG
$4,630.00

$301.00


Available to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
$301.00
100 G
$695.00
250 G
$1,670.00
1 KG
$4,630.00

About This Item

Linear Formula:
AgNO3
CAS Number:
Molecular Weight:
169.87
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-853-9
NACRES:
NA.21
grade:
ACS reagent
form:
crystals
solid

$301.00


Available to ship on31 March 2025Details


Request a Bulk Order

grade

ACS reagent

Quality Level

Agency

reag. ISO
reag. Ph. Eur.

vapor density

5.8 (vs air)

product line

EMSURE®

Assay

99.8-100.5% (silver nitrate titration)

form

crystals
solid

impurities

≤0.01% Substances not precipitated by hydrochloric acid (ACS)
≤0.01% Substances not precipitated by hydrochloric acid (ISO)
≤0.3% Substances not precipitated by hydrochloric acid (Reag Ph Eur)

pH

5.4-6.4 (20 °C, 100 g/L in H2O)

bp

444 °C/1013 hPa (decomposition)

mp

212 °C (dec.) (lit.)

density

4.35 g/cm3 at 20 °C

bulk density

2350 kg/m3

anion traces

chloride (Cl-): ≤0.0005%
sulfate (SO42-): ≤0.002%

cation traces

Cd: ≤0.0001%
Cu: ≤0.0002%
Fe: ≤0.0002%
Mn: ≤0.0005%
Ni: ≤0.0005%
Pb: ≤0.001%
Tl: ≤0.001%
Zn: ≤0.0001%

storage temp.

no temp limit

SMILES string

[O-][N+]([O-])=O.[Ag+]

InChI

1S/Ag.NO3/c;2-1(3)4/q+1;-1

InChI key

SQGYOTSLMSWVJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Fluoride-Ion-Responsive Sol-Gel Transition in an L-Cysteine/AgNO3 System: Self-Assembly Peculiarities and Anticancer Activity.: This research presents a novel use of silver nitrate in a sol-gel system that responds to fluoride ions. The findings contribute to the development of responsive materials with potential applications in drug delivery systems and tissue engineering, showcasing the versatility of silver nitrate in advanced material science (Vishnevetskii et al., 2024).

Analysis Note

Assay (argentometric): 99.8 - 100.5 %


Identity: passes test


Acidity or alkalinity: passes test


Free acid: passes test


Appearance of solution: passes test


Chloride (Cl): ≤ 0.0005 %


Sulfate (SO₄): ≤ 0.002 %


Cd (Cadmium): ≤ 0.0001 %


Cu (Copper): ≤ 0.0002 %


Fe (Iron): ≤ 0.0002 %


Mn (Manganese): ≤ 0.0005 %


Ni (Nickel): ≤ 0.0005 %


Pb (Lead): ≤ 0.001 %


Tl (Thallium): ≤ 0.001 %


Zn (Zinc): ≤ 0.0001 %


Copper and substances precipitated by NH₃ (Al, Bi, Cu, Pb): passes test


Substances not precipitated by hydrochloric acid (Reag Ph Eur): ≤ 0.3 %
Substances not precipitated by hydrochloric acid (ACS): ≤ 0.01 %
Substances not precipitated by hydrochloric acid (ISO): ≤ 0.01 %
Corresponds to ACS, ISO, Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Ox. Sol. 2 - Repr. 1B - Skin Corr. 1A

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

EMSURE® Grade Inorganic Solvents and Reagents offer exceptional purity and quality control, ensuring reliable, compliant results for regulated analyses.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service