Skip to Content
Merck
All Photos(1)

Key Documents

1.01310

Millipore

Brilliant green (hydrogen sulfate) (C.I. 42040)

suitable for microbiology

Synonym(s):

Brilliant green (hydrogen sulfate) (C.I. 42040), Diamond green G, Solid green, Ethyl green

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$157.00

$157.00


Estimated to ship on02 June 2025


Request more information

Select a Size

Change View
50 G
$157.00

About This Item

Empirical Formula (Hill Notation):
C27H34N2O4S
CAS Number:
Molecular Weight:
482.63
MDL number:
UNSPSC Code:
41106212
EC Index Number:
211-190-1
NACRES:
NA.74

$157.00


Estimated to ship on02 June 2025


Request more information

potency

300-400 mg/kg LD50, oral (Rat)

Quality Level

pH

2.5 (25 °C, 10 g/L in H2O)

mp

210 °C (decomposition)

solubility

100 g/L

bulk density

500‑600 kg/m3

application(s)

microbiology

storage temp.

2-30°C

SMILES string

[S](=O)(=O)([O-])O.[N+](=C1C=CC(=C(c3ccc(cc3)N(CC)CC)c2ccccc2)C=C1)(CC)CC

InChI

1S/C27H33N2.H2O4S/c1-5-28(6-2)25-18-14-23(15-19-25)27(22-12-10-9-11-13-22)24-16-20-26(21-17-24)29(7-3)8-4;1-5(2,3)4/h9-21H,5-8H2,1-4H3;(H2,1,2,3,4)/q+1;/p-1

InChI key

NNBFNNNWANBMTI-UHFFFAOYSA-M

Analysis Note

Dye content (spectrophotometrically): ≥ 80 %
Identity (UV/VIS-Spectrum): passes test
Absorption maximum λmax. (ethanol 50 %): 628 - 632 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.003 g/l; ethanol 50 %): 1860 - 2370
TLC-Test: passes test
Loss on drying (110 °C): ≤ 5 %
Suitability for microbiology: passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service