1.01236
Agarose
for analytical nucleic acid electrophoresis
Synonym(s):
Agarose
Select a Size
Select a Size
About This Item
Recommended Products
form
solid
mp
60-90 °C
bulk density
700 kg/m3
foreign activity
DNAses (Exo- and endonucleases), none detected
RNAse, none detected
storage temp.
15-25°C
SMILES string
O1[C@H]([C@@H]([C@H]([C@H]([C@H]1CO)O)O[C@@H]4O[C@@H]5[C@H]([C@@H](OC5)[C@@H]4O)O[C@@H]6O[C@@H]([C@@H]([C@@H]([C@H]6O)O)O)CO)O)O[C@H]2[C@H]3OC[C@@H]2O[C@H]([C@H]3O)O
InChI
1S/C24H38O19/c25-1-5-9(27)11(29)12(30)22(38-5)41-17-8-4-36-20(17)15(33)24(40-8)43-18-10(28)6(2-26)39-23(14(18)32)42-16-7-3-35-19(16)13(31)21(34)37-7/h5-34H,1-4H2/t5-,6-,7+,8+,9+,10+,11+,12-,13+,14-,15+,16-,17-,18+,19+,20+,21-,22+,23+,24+/m1/s1
InChI key
MJQHZNBUODTQTK-WKGBVCLCSA-N
Related Categories
Analysis Note
Appearance (description): crystalline powder
Electroendoosmosis: 0.05 - 0.13
Gel point: 34 - 38 °C
Sulphur (as SO₄): ≤ 0.14 %
Sulfated ash (800 °C): ≤ 0.4 %
Loss on drying (105 °C, 2 h): ≤ 7.0 %
Performance test (Nucleic acid electrophoresis): passes test
DNases (Exo- and endonucleases): not detectable
RNases: not detectable
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Learn how water impurities can affect DNA and RNA electrophoresis and blotting. High purity water purified using ultrafiltration is suitable for nucleic acid electrophoresis, Northern and Southern blotting.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service