Skip to Content
Merck
All Photos(1)

Key Documents

1.01145

Supelco

Ammonium chloride

for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Synonym(s):

Ammonium chloride, Sal ammoniac

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$98.60
1 KG
$152.00
5 KG
$553.00
25 KG
$3,030.00
50 KG
$4,920.00

$98.60


Available to ship on07 April 2025Details


Request a Bulk Order

Select a Size

Change View
500 G
$98.60
1 KG
$152.00
5 KG
$553.00
25 KG
$3,030.00
50 KG
$4,920.00

About This Item

Empirical Formula (Hill Notation):
H4ClN
CAS Number:
Molecular Weight:
53.49
MDL number:
UNSPSC Code:
12352301
EC Index Number:
235-186-4
NACRES:
NA.21
grade:
ACS reagent
form:
crystalline powder
solubility:
372 g/L (External MSDS)

$98.60


Available to ship on07 April 2025Details


Request a Bulk Order

grade

ACS reagent

Quality Level

Agency

reag. ISO
reag. Ph. Eur.

vapor pressure

1.3 hPa ( 30 °C)

product line

EMSURE®

Assay

≥99.8% (silver nitrate titration)
99.0-100.5% dry basis (silver nitrate titration)

form

crystalline powder

autoignition temp.

>400 °C

potency

1410 mg/kg LD50, oral (Rat)

impurities

≤0.005% Insoluble matter

ign. residue

≤0.01% (as sulfate)

loss

≤1% loss on drying, 105°C

pH

4.7 (25 °C, 200 g/L in H2O, External MSDS)

mp

338 °C (sublimed; External MSDS)

solubility

372 g/L (External MSDS)

density

1.53 g/cm3 at 25 °C

bulk density

600‑900 kg/m3

anion traces

bromide (Br-): passes test
iodide (I-): passes test
nitrate (NO3-): ≤0.0005%
phosphate (PO43-): ≤0.0002%
sulfate (SO42-): ≤0.002%

cation traces

Ca: ≤0.0005%
Cu: ≤0.0002%
Fe: ≤0.0002%
K: ≤0.005%
Mg: ≤0.0005%
Na: ≤0.005%
Ni: ≤0.0001%
Pb: ≤0.0001%
Zn: ≤0.0002%
heavy metals: ≤0.0005%

storage temp.

2-30°C

SMILES string

[Cl-].N.[H+]

InChI

1S/ClH.H3N/h1H;1H3

InChI key

NLXLAEXVIDQMFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Restoration of the soil fertility under Cr(VI) and artificial drought condition by the utilization of plant growth-promoting Bacillus spp. SSAU2.: The study investigates the role of ammonium chloride in enhancing the effectiveness of Bacillus spp. in soil fertility restoration under stress conditions, showcasing its relevance in environmental science and microbiology (Sharma et al., 2024).


Analysis Note

Assay (argentometric): ≥ 99.8 %
Assay (argentometric; calculated on dried substance): 99.0 - 100.5 %
Identity: passes test
Appearance of solution: passes test
Insoluble matter: ≤ 0.005 %
Acidity or alkalinity: passes test
pH-value (5 %; water, 25 °C): 4.5 - 5.5
Bromide, Iodide (Br,I): passes test
Nitrate (NO₃): ≤ 0.0005 %
Phosphate (PO₄): ≤ 0.0002 %
Sulfate (SO₄): ≤ 0.002 %
Heavy metals (ACS): ≤ 0.0005 %
Ca (Calcium): ≤ 0.0005 %
Cu (Copper): ≤ 0.0002 %
Fe (Iron): ≤ 0.0002 %
K (Potassium): ≤ 0.005 %
Mg (Magnesium): ≤ 0.0005 %
Na (Sodium): ≤ 0.005 %
Ni (Nickel): ≤ 0.0001 %
Pb (Lead): ≤ 0.0001 %
Zn (Zinc): ≤ 0.0002 %
Loss on Drying (105°C): ≤ 1.0 %
Residue on ignition (as sulfate): ≤ 0.01 %
Corresponds to ACS,ISO,Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service