Skip to Content
Merck
All Photos(1)

Key Documents

1.01134

Supelco

Ammonium carbamate

for analysis EMSURE®

Synonym(s):

Ammonium carbamate, Ammonium carbaminate, Carbamic acid ammonium salt

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$198.00

$198.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
500 G
$198.00

About This Item

Empirical Formula (Hill Notation):
CH6N2O2
CAS Number:
Molecular Weight:
78.07
MDL number:
UNSPSC Code:
12352107
EC Index Number:
214-185-2
NACRES:
NA.21
form:
solid
solubility:
790 g/L

$198.00


Estimated to ship on30 May 2025


Request a Bulk Order

vapor pressure

82 hPa ( 20 °C)

Quality Level

product line

EMSURE®

Assay

≥99.5% (alkalimetric)

form

solid

potency

681-1470 mg/kg LD50, oral (Rat)

ign. residue

≤0.002% (as sulfate)

pH

10 (20 °C, 100 g/L in H2O)

mp

152 °C (closed capillary tube)

solubility

790 g/L

density

1.6 g/cm3 at 20 °C

bulk density

750 kg/m3

anion traces

chloride (Cl-): ≤0.0003%
nitrate (NO3-): ≤0.001%
phosphate (PO43-): ≤0.0005%
sulfate (SO42-): ≤0.001%

cation traces

Ca: ≤0.003%
Fe: ≤0.0002%
heavy metals (as Pb): ≤0.0005%

storage temp.

15-25°C

SMILES string

[N+H4].NC(=O)[O-].O

InChI

1S/CH3NO2.H3N.H2O/c2-1(3)4;;/h2H2,(H,3,4);1H3;1H2

InChI key

SFWLALIEAXEULE-UHFFFAOYSA-N

Application


  • Oxalic acid as a dual C1 surrogate for heterogeneous palladium-catalyzed tandem four-component quinazolinone synthesis.: This study explores the use of ammonium carbamate in synthesizing quinazolinones via a novel palladium-catalyzed method, highlighting its application in organic synthesis (Sheetal et al., 2024).

  • High-Capacity, Cooperative CO(2) Capture in a Diamine-Appended Metal-Organic Framework through a Combined Chemisorptive and Physisorptive Mechanism.: Ammonium carbamate is discussed in the context of enhancing CO2 capture efficiency in metal-organic frameworks, showcasing its potential in environmental applications (Zhu et al., 2024).

  • Thermal Synthesis of Carbamic Acid and Its Dimer in Interstellar Ices: A Reservoir of Interstellar Amino Acids.: The paper investigates the formation of carbamic acid from ammonium carbamate under interstellar conditions, providing insights into astrochemistry and the origins of amino acids in space (Marks et al., 2023).


Analysis Note

Assay (alkalimetric): ≥ 99.5 %
Chloride (Cl): ≤ 0.0003 %
Nitrate (NO₃): ≤ 0.001 %
Phosphate (PO₄): ≤ 0.0005 %
Sulfate (SO₄): ≤ 0.001 %
Heavy metals (as Pb): ≤ 0.0005 %
Ca (Calcium): ≤ 0.003 %
Fe (Iron): ≤ 0.0002 %
Residue on ignition (as sulfate): ≤ 0.002 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service