Skip to Content
Merck
All Photos(1)

Key Documents

1.00898

Millipore

E.coli / Coliform Selective-Supplement

for Escherichia coli, for coliforms, pkg of 10 vials, for use with ChromoCult® Coliform Agar

Synonym(s):

E. coli / Coliform supplement, E. coli / Coliform antibiotic Selective Supplement

Sign Into View Organizational & Contract Pricing

Select a Size

10 VIAL
$215.00

$215.00


Estimated to ship on28 May 2025


Request more information

Select a Size

Change View
10 VIAL
$215.00

About This Item

UNSPSC Code:
41106212
NACRES:
NA.74

$215.00


Estimated to ship on28 May 2025


Request more information

form

lyophilized powder

Quality Level

composition

Cefsulodine, 2.5 mg per vial
Vancomycin, 2.5 mg per vial

packaging

pkg of 10 vials

technique(s)

microbiological culture: suitable

pH

3.5-4.5 (25 °C, 14 g/L in H2O)

application(s)

food and beverages
microbiology

compatibility

for use with ChromoCult® Coliform Agar

storage temp.

2-8°C

suitability

Escherichia coli
coliforms

General description

E. coli / Coliform supplement is a mixture of two antibiotics in lyophilized form. Vancomycin inhibits the growth of Gram-positive bacteria. Cefsulodin suppresses the growth of Pseudomonas spp. and Aeromonas spp. This selective supplement can be used in combination with most E. coli / Coliform enrichment media and plating agars.

Application

E. coli / Coliform Selective-Supplement is an additive for the preparation of selective culture media for the detection of E. coli / Coliforms in water, food, and environmental samples.

Linkage

Replaces: 1.00898

Analysis Note

Supplement tested in Chromocult Coliform agar art. no. 1.10426.
Recovery on test medium
- Escherichia coli DSM 502:
≥ 70 %
- Citrobacter freundii ATCC 8090: ≥ 70 %
Growth
- Aeromonas hydrophila ATCC 7966 (WDCM 00063):
inhibited
- Pseudomonas aeruginosa ATCC 27853 (WDCM 00025): inhibited
- Bacillus licheniformis ATCC 14580 (WDCM 00068): inhibited
- Staphylococcus aureus ATCC 6538 (WDCM 00032): inhibited
Colony colour
- Escherichia coli DSM 502:
blue
- Citrobacter freundii ATCC 8090: red
Incubation: 24 hrs.; 35 °C; aerobic.

Legal Information

CHROMOCULT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service