Skip to Content
Merck
All Photos(1)

Key Documents

1.00761

Supelco

Sulfurous acid

5-6% SO₂, for analysis EMSURE®

Synonym(s):

Sulfur dioxide solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$111.00
2.5 L
$218.00

$111.00


Available to ship on02 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 L
$111.00
2.5 L
$218.00

About This Item

UNSPSC Code:
12352301
NACRES:
NA.21

$111.00


Available to ship on02 April 2025Details


Request a Bulk Order

Product Name

Sulfurous acid 5-6% SO₂, for analysis EMSURE®

grade

reagent

Quality Level

product line

EMSURE®

form

liquid

concentration

5-6% (H2SO3)

impurities

≤0.02 ppm Ag (Silver)
≤0.02 ppm Be (Beryllium)
≤0.02 ppm Co (Cobalt)
≤0.02 ppm Cr (Chromium)
≤0.02 ppm Cu (Copper)
≤0.02 ppm Li (Lithium)
≤0.02 ppm Mn (Manganese)
≤0.02 ppm Mo (Molybdenum)
≤0.02 ppm Ni (Nickel)
≤0.02 ppm Pb (Lead)
≤0.02 ppm Sr (Strontium)
≤0.05 ppm Al (Aluminium)
≤0.05 ppm Ba (Barium)
≤0.05 ppm Cd (Cadmium)
≤0.05 ppm Ge (Germanium)
≤0.05 ppm Tl (Thallium)
≤0.05 ppm V (Vanadium)
≤0.05 ppm Zn (Zinc)
≤0.1 ppm Bi (Bismuth)
≤0.1 ppm K (Potassium)
≤0.1 ppm Mg (Magnesium)
≤0.1 ppm Ti (Titanium)
≤0.1 ppm Zr (Zirconium)
≤0.5 ppm As (Arsenic)
≤0.5 ppm Fe (Iron)
≤0.5 ppm Na (Sodium)
≤1 ppm Ca (Calcium)
≤5 ppm Chloride (Cl)

ign. residue

≤0.002% (as sulfate)

density

1.03 g/cm3 at 20 °C

storage temp.

15-25°C

Related Categories

Analysis Note

Assay (iodometric, SO₂): 5 - 6 %
Chloride (Cl): ≤ 5 ppm
Ag (Silver): ≤ 0.02 ppm
Al (Aluminium): ≤ 0.05 ppm
As (Arsenic): ≤ 0.5 ppm
Ba (Barium): ≤ 0.05 ppm
Be (Beryllium): ≤ 0.02 ppm
Bi (Bismuth): ≤ 0.1 ppm
Ca (Calcium): ≤ 1 ppm
Cd (Cadmium): ≤ 0.05 ppm
Co (Cobalt): ≤ 0.02 ppm
Cr (Chromium): ≤ 0.02 ppm
Cu (Copper): ≤ 0.02 ppm
Fe (Iron): ≤ 0.5 ppm
Ge (Germanium): ≤ 0.05 ppm
K (Potassium): ≤ 0.1 ppm
Li (Lithium): ≤ 0.02 ppm
Mg (Magnesium): ≤ 0.1 ppm
Mn (Manganese): ≤ 0.02 ppm
Mo (Molybdenum): ≤ 0.02 ppm
Na (Sodium): ≤ 0.5 ppm
Ni (Nickel): ≤ 0.02 ppm
Pb (Lead): ≤ 0.02 ppm
Sr (Strontium): ≤ 0.02 ppm
Ti (Titanium): ≤ 0.1 ppm
Tl (Thallium): ≤ 0.05 ppm
V (Vanadium): ≤ 0.05 ppm
Zn (Zinc): ≤ 0.05 ppm
Zr (Zirconium): ≤ 0.1 ppm
Residue on ignition (as sulfate): ≤ 0.002 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service