Skip to Content
Merck
All Photos(1)

Key Documents

1.00684

Supelco

Sulfanilic acid

for elemental analysis

Synonym(s):

Sulfanilic acid, 4-Aminobenzenesulfonic acid, Aniline-4-sulfonic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$117.00

$117.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
$117.00

About This Item

Empirical Formula (Hill Notation):
C6H7NO3S
CAS Number:
Molecular Weight:
173.19
MDL number:
UNSPSC Code:
12352106
EC Index Number:
204-482-5
NACRES:
NA.21

$117.00


Estimated to ship on31 May 2025


Request a Bulk Order

grade

for elemental analysis

Quality Level

form

solid

autoignition temp.

>400 °C

potency

12300 mg/kg LD50, oral (Rat)

pH

2.5 (20 °C, 10 g/L in H2O)

mp

288 °C (decomposes)

solubility

10 g/L

density

1.485 g/cm3 at 25 °C

bulk density

620 kg/m3

storage temp.

15-25°C

SMILES string

[S](=O)(=O)(O)c1ccc(cc1)N

InChI

1S/C6H7NO3S/c7-5-1-3-6(4-2-5)11(8,9)10/h1-4H,7H2,(H,8,9,10)

InChI key

HVBSAKJJOYLTQU-UHFFFAOYSA-N

General description

for elemental analysis

Application

Sulfanilic acid standard for elemental analysis for analysis. CAS 121-57-3, EC Number 204-482-5, chemical formula NH₂C₆H₄SO₃H.

Analysis Note

Identity (IR-spectrum)conformsC content (mean value fivefold determination)41.3 - 41.8 %H content (mean value fivefold determination)3.9 - 4.3 %N-content (mean value fivefold determination)7.7 - 8.3 %O content (mean value fivefold determination)27.5 - 28.5 %S content (mean value fivefold determination)18.2 - 18.8 %Water (according to Karl Fischer)≤ 0.5 %Suitable for determination of C,H,N,O,S

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service