Skip to Content
Merck
All Photos(1)

Key Documents

1.00617

Supelco

Sulfate Test, photometric

photometric, 50-500 mg/L (SO42-), Spectroquant®

Sign Into View Organizational & Contract Pricing

Select a Size

25 TESTS
$280.00

$280.00


Available to ship on31 March 2025Details


Request a Bulk OrderRequest more information

Select a Size

Change View
25 TESTS
$280.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

$280.00


Available to ship on31 March 2025Details


Request a Bulk OrderRequest more information

Product Name

Sulfate Cell Test, photometric, 50-500 mg/L (SO42-), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 25 tests

specific analyte(s)

sulfate

measuring range

50-500 mg/L (SO42-)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Barium sulfate, turbidimetric)

storage temp.

15-25°C

General description

Method: photometric 50 - 500 mg/l SO₄²⁻ Spectroquant®
The method is analogous to EPA 375.4, APHA 4500-SO4 2- E, and ASTM D516-11.
This Spectroquant® Sulfate Cell Test allows the accurate quantification of the sulfate content in aqueous samples.
Method applied: Sulfate ions react with barium ions to form slightly soluble barium sulfate. The resulting turbidity is measured photometrically (turbidimetric method).
The Spectroquant® Cell Tests come with prefilled 16 mm round cells and all the required reagents to perform the analysis according to the instruction leaflet provided.
All Spectroquant® Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Met. Corr. 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luis A Cueto et al.
Environmental science and pollution research international, 27(14), 15984-15993 (2019-05-06)
Phosphorus (P) is a nutrient for plant growth but also a pollutant in water bodies causing eutrophication. The source of P is mainly human and animal wastewater and runoffs from different land uses. The objective of the present study is

Protocols

Photometric or turbidimetric determination subsequent to water extraction/fusion melting

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service