Skip to Content
Merck
All Photos(1)

Documents

1.00614

Supelco

Nitrate Test, photometric

photometric, 23-255 mg/L (NO3-N), 102-996 mg/L (NO3-), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Nitrate Cell Test, photometric, 23-255 mg/L (NO3-N), 102-996 mg/L (NO3-), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 25 tests

specific analyte(s)

nitrate

measuring range

102-996 mg/L (NO3-)
23-255 mg/L (NO3-N)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (2,6-Dimethylphenol)

storage temp.

15-25°C

General description

Method: photometric, DMP 23 - 225 mg/l, NO3-N 102 - 996 mg/l NO3- Spectroquant®

The method corresponds to ISO 23696-1, a norm standardizing the use of this "small-scale sealed tube method" for this important parameter. The method is also analogous to DIN 38405-9.

This Spectroquant® Nitrate Cell Test allows the accurate quantification of nitrate in water samples. Nitrate is an essential nutrient required for the growth of animals and plants. However, elevated levels of nitrate can be a health risk, in particular for pregnant women and infants. Therefore, nitrate levels in water sources are regulated in many countries.

Further fields of applications are the analysis of foods & beverages (e.g. whey) and of environmental and agricultural samples (e.g. soil).

The Spectroquant® Cell Tests come with prefilled 16 mm round cells and all the required reagents to perform the analysis according to the instruction leaflet provided.

All Spectroquant® Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Application


  • Application of a nitrifying bacterium Pseudomonas sp. HIB_D to reduce nitrogen waste in the Litopenaeus vannamei cultivation environment.: Explores the use of nitrate cell tests in aquaculture to monitor and manage nitrogen levels, enhancing environmental sustainability and shrimp health (Hastuti et al., 2023).

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service