Skip to Content
Merck
All Photos(1)

Key Documents

1.00255

Supelco

Diphenylamine-4-sulfonic acid barium salt

redox indicator

Synonym(s):

Diphenylamine-4-sulfonic acid barium salt, 4-Anilinobenzene sulfonic acid barium salt, Barium diphenylaminesulfonate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$248.00

$248.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
$248.00

About This Item

Empirical Formula (Hill Notation):
C24H20BaN2O6S2
CAS Number:
Molecular Weight:
633.88
MDL number:
UNSPSC Code:
12352107
EC Index Number:
228-278-0
NACRES:
NA.21

$248.00


Estimated to ship on31 May 2025


Request a Bulk Order

Product Name

Diphenylamine-4-sulfonic acid barium salt, redox indicator

Quality Level

form

solid

technique(s)

titration: suitable

loss

≤5% loss on drying, 105°C

pH

5.9 (20 °C, 10 g/L in H2O, slurry)

mp

300 °C

solubility

water: soluble at 20 °C

bulk density

220 kg/m3

λmax

290-295 nm in water

storage temp.

2-30°C

SMILES string

[Ba+2].[S](=O)(=O)([O-])c3ccc(cc3)Nc4ccccc4.[S](=O)(=O)([O-])c1ccc(cc1)Nc2ccccc2

InChI

1S/2C12H11NO3S.Ba/c2*14-17(15,16)12-8-6-11(7-9-12)13-10-4-2-1-3-5-10;/h2*1-9,13H,(H,14,15,16);/q;;+2/p-2

InChI key

IVCNVXFNTKXMCA-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Identity (IR-spectrum): passes test
Absorption maximum λmax. (water): 290 - 295 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.001 %; water; calculated on dried substance): 600 - 700
Loss on Drying (105 °C): ≤ 5 %
Suitability as redox indicator: passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service