Skip to Content
Merck
All Photos(1)

Key Documents

1.00251

Sigma-Aldrich

Reticulin silver plating kit acc. to Gordon & Sweets

suitable for histology

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
$554.00

$554.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
1 SET
$554.00

About This Item

UNSPSC Code:
41116130
NACRES:
NA.41

$554.00


Estimated to ship on29 May 2025


Request a Bulk Order

IVD

for in vitro diagnostic use

Quality Level

dilution

(for the detection of reticular fibers)

application(s)

clinical testing
diagnostic assay manufacturing
hematology

histology

storage temp.

2-8°C

General description

The Reticulin silver plating kit according to Gordon & Sweets -for the detection of reticular fibers in histological tissue specimens, can be used to visualize human reticular fibers, which are components of membrane structures, capillaries and peripheral nerve fibers, as well as organs such as the liver, kidneys and hematopoetic organs. Fibers that can not be adequately visualized using an H&E stain may be rendered visible through a silver plating stain. Reticulin visualization with the silver stain acc. to Gordon and Sweets gives greater contrast and sharpness against the background. Gordon & Sweets′ silver stain for reticulin may be followed by nuclear staining with nuclear fast red-aluminum sulfate solution 0.1 % (Product number 1.00121) when nuclei, collagen and the background are to be stained in addition to the reticular fibers.
The kit is sufficient for up to 110 section slides. It is an IVD registered product and is CE certified, thus, it can be used in an accredited laboratory. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Analysis Note

Suitability for microscopy: passes test
reticular fibers: black
Nuclei: red
Collagen: red
Background: red

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Irrit. 2 - Met. Corr. 1 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service