Skip to Content
Merck
All Photos(1)

Key Documents

100106

Sigma-Aldrich

A23187, Mixed Calcium-Magnesium Salt

Useful in cell activation experiments when calcium dose-response data are not required.

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$264.00

$264.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$264.00

About This Item

UNSPSC Code:
51111800
NACRES:
NA.77

$264.00


Estimated to ship on25 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (TLC)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: soluble
ethanol: soluble

shipped in

ambient

storage temp.

2-8°C

General description

Useful in cell activation experiments when calcium dose-response data are not required.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Useful in cell activation experiments when calcium dose-response data are not required
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Preparation Note

For use in aqueous systems, preparation of a 10 mM stock in DMSO is recommended. Microliter amounts of this solution can be added to H₂O with rapid mixing (e.g. 10 µl into 5 ml of H₂O). In aqueous systems, the ionophore aggregates over time.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 2 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service