Skip to Content
Merck
All Photos(2)

Key Documents

09-827

Sigma-Aldrich

Anti-HDAC11 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

histone deacetylase 11

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$869.00

$869.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$869.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$869.00


Estimated to ship on26 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse, rat

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

HDAC11 is responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3 and H4). Histone deacetylation is a marker for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes. The predominantly nuclear HDAC11, which interacts with HDAC6, is weakly expressed in most tissues, and strongly expressed in brain, heart, skeletal muscle, kidney and testis. Its activity is inhibited by trapoxin, a known histone deacetylase inhibitor.

Specificity

This antibody recognizes HDAC11.

Immunogen

Epitope: Amino Acid: 228-246
Linear Peptide corresponding to human HDAC11 at amino acids 228-246.

Application

Anti-HDAC11 Antibody is a Rabbit Polyclonal Antibody for detection of HDAC11 also known as histone deacetylase 11 & has been validated in WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histone Modifying Proteins

Cell Cycle, DNA Replication & Repair
Western Blot (SNAP ID) Analysis: 0.5 µg/ml from a representative lot detected HDAC11 on 10 µg of human brain tissue lysate.

Quality

Evaluated by Western Blot in human brain tissue lysate.

Western Blot Analysis: 0.05 µg/ml of this antibody detected HDAC11 on 10 µg of human brain tissue lysate.

Target description

39 kDa

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human brain tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service