Skip to Content
Merck
All Photos(1)

Key Documents

09-823

Sigma-Aldrich

Anti-JMJD1A Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

JMJD, lysine (K)-specific demethylase 3A, testis-specific protein A, jumonji domain containing 1A, jumonji C domain-containing histone demethylase 2A, jumonji domain containing 1, Jumonji domain-containing protein 1A, JmjC domain-containing histone demet

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$739.00

$739.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$739.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$739.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

technique(s)

ChIP: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KDM3A(55818)

General description

JMJD1A (jumonji domain-containing 1a) is a histone demethylase that plays a key role in the histone code. Functionally, this protein is known to demethylate mono- and dimethylated histoneH3 at lysine 9, while demonstrating virtually no demethylation activity for trimethylated lysine 9 of histone H3. A product of the JHDM2A gene, this protein has been demonstrated to play a role in hypoxia, spermatogenesis, and cell differentiation with loss of function of JMJD1A resulting embryonic stem cell differentiation.

Immunogen

GST-tagged recombinant protein corresponding to mouse JMJD1A.

Application

Immunoprecipitation Analysis: 5 µg of a representative lot of this antibody immunoprecipitated JMJD1A in HeLa nuclear extract.

Western Blot Analysis: A representitive lot of this antibody detected JMJD1A in mouse ESC chromatin extract.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histone Modifying Proteins
Use Anti-JMJD1A Antibody (Rabbit Polyclonal Antibody) validated in WB, IP to detect JMJD1A also known as lysine (K)-specific demethylase 3A, testis-specific protein A.

Quality

Evaluated by Western Blot in HEK293 nuclear extract.

Western Blot Analysis: 1 µg/mL of this antibody detected JMJD1A on 10 µg of HEK293 nuclear extract.

Target description

~147 kDa observed.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HEK293 nuclear extract.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glauber R de S Araújo et al.
Methods in molecular biology (Clifton, N.J.), 2775, 141-153 (2024-05-17)
This chapter describes methodological details for preparing specimens of Cryptococcus neoformans (although it can be applied to any species of the genus) and their subsequent analysis by scanning and transmission electron microscopy. Adaptations to conventional protocols for better preservation of
Alexander Malogolovkin et al.
Journal of medical virology, 95(1), e28252-e28252 (2022-10-23)
Zika virus (ZIKV) is one of several examples of an unprecedented pandemic spread and against which there is currently no suitable vaccine or treatment. Here, we constructed and characterized recombinant baculovirus-derived ZIKV-like particles (Zika VLPs) to study ZIKV-antibody interactions. These VLPs
Gina La Sala et al.
Journal of neuroscience research (2020-12-23)
Mammalian cerebellar astrocytes critically regulate the differentiation and maturation of neuronal Purkinje cells and granule precursors. The G protein-coupled receptor 37-like 1 (Gpr37l1) is expressed by Bergmann astrocytes and interacts with patched 1 (Ptch1) at peri-ciliary membranes. Cerebellar primary astrocyte
Itze C Navarro-Hernandez et al.
The FEBS journal, 287(16), 3449-3471 (2020-01-21)
B lymphocytes are a leukocyte subset capable of developing several functions apart from differentiating into antibody-secreting cells. These processes are triggered by external activation signals that induce changes in the plasma membrane properties, regulated by the formation of different lipid-bilayer

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service