Skip to Content
Merck
All Photos(2)

Key Documents

09-466

Sigma-Aldrich

Anti-phospho-CrkL (Tyr207) Antibody

Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$683.00

$683.00


Estimated to ship on07 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$683.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$683.00


Estimated to ship on07 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat, human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pTyr207)

Gene Information

human ... CRKL(1399)

General description

CRKL belongs to a family of adaptor proteins containing SH2/SH3 domains that couple to effector proteins including p130CAS, SOS, Abl, and Arg. CRKL plays a role in the mediation of Integrin- and Rac-dependent cell migration and in Ras-dependent activation of JNK. The protein is also involved in T-cell activation. Phosphorylation of CrkL on Tyr207 is known to be correlated with its inhibition.

Specificity

Recognizes CrkL only when phosphorylated on Tyr207.

Immunogen

KLH synthetic peptide including and encompassing Tyr 207 of human CrkL.

Application

Detect phospho-CrkL (Tyr207) using this Anti-phospho-CrkL (Tyr207) Antibody validated for use in WB.
Research Category
Signaling
Research Sub Category
Cytoskeletal Signaling

Quality

Routinely evaluated by western blot on K563 cell lysates.

Target description

39 kDa

Physical form

Purified rabbit polyclonal IgG in storage buffer containing PBS with 0.05% sodium azide in 50% glycerol.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
K562

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service