Skip to Content
Merck
All Photos(1)

Key Documents

07-833

Sigma-Aldrich

Anti-phospho-Gab1 (Tyr627) Antibody

Upstate®, from rabbit

Synonym(s):

GAB1, GRB2-associated binding protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pTyr627)

Gene Information

mouse ... Gab1(14388)

Specificity

Gab1 phosphorylated on Tyrosine residue 627.
Predicted to cross-react with human based on 100% sequence homology, but have not been tested.

Immunogen

Peptide corresponding to amino acid region encompassing the human and mouse phospho-Gab1 (Tyr627)

Application

Immunoblot Analysis: 1:1000 dilutions of this lot detected phospho-Gab1 (Tyr627) in RIPA lysate of PDGF treated NIH3T3 cells.
Research Category
Signaling
Research Sub Category
MAP Kinases
This Anti-phospho-Gab1 (Tyr627) Antibody is validated for use in WB for the detection of phospho-Gab1 (Tyr627).

Quality

Routinely evaluated by immunoblot.

Target description

115 kDa

Physical form

100 μL of affinity purified rabbit polyclonal IgG in 50% storage buffer (PBS (without Mg2+ and Ca2+), pH 7.3 containing 1.0 mg/mL BSA (IgG, protease free) and 0.05% sodium azide) and 50% glycerol.
Affinity purified

Storage and Stability

Stable for 2 years at -20°C from date of shipment. Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variability in freezer temperatures below -20°C may cause glycerol-containing solutions to become frozen during storage.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Shinohara et al.
The Journal of biological chemistry, 276(22), 18941-18946 (2001-03-23)
Gab-1 is a multiple docking protein that is tyrosine phosphorylated by receptor tyrosine kinases such as c-Met, hepatocyte growth factor/scatter factor receptor, and epidermal growth factor receptor. We have now demonstrated that cell-cell adhesion also induces marked tyrosine phosphorylation of
C Boccaccio et al.
Nature, 391(6664), 285-288 (1998-01-24)
Hepatocyte growth factor (HGF) induces a three-phase response leading to the formation of branched tubular structures in epithelial cells. The HGF receptor tyrosine kinase works through a Src homology (SH2) docking site that can activate several signalling pathways. The first
Lilly Y W Bourguignon et al.
The Journal of biological chemistry, 278(32), 29420-29434 (2003-05-16)
In this study we have examined CD44 (a hyaluronan (HA) receptor) interaction with a RhoA-specific guanine nucleotide exchange factor (p115RhoGEF) in human metastatic breast tumor cells (MDA-MB-231 cell line). Immunoprecipitation and immunoblot analyses indicate that both CD44 and p115RhoGEF are
M Holgado-Madruga et al.
Nature, 379(6565), 560-564 (1996-02-08)
The protein Grb2 plays a central role in signalling by receptor protein-tyrosine kinases, where its SH2 domain binds to the receptor and its two SH3 domains link to effectors. One target effector is Sos, so Grb2 links receptor protein-tyrosine kinases

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service