Skip to Content
Merck
All Photos(3)

Key Documents

07-369-I

Sigma-Aldrich

Anti-dimethyl Histone H3 Antibody (Lys36)

serum, from rabbit

Synonym(s):

Histone H3.1t, H3/t/ H3t/ H3/g

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$922.00

$922.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$922.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$922.00


Estimated to ship on26 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

species reactivity (predicted by homology)

vertebrates (based on 100% sequence homology)

technique(s)

ChIP: suitable
dot blot: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

dimethylation (Lys36)

Gene Information

human ... H3C1(8350)

General description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.

Immunogen

KLH-conjugated linear peptide corresponding to a region near the N-terminus of human dimethyl Histone H3 methylated at Lys36.

Application

Anti-dimethyl Histone H3 Antibody (Lys36) is a highly specific rabbit polyclonal antibody, that targets Histone H3 & has been tested in western blotting, Dot Blot & Chromatin IP (ChIP).
Dot Blot Analysis: A 1:10,000 dilution from a representative lot detected dimethyl Histone H3 (Lys36), but not unmethylated Histone H3 or other peptides corresponding to modified histones.

Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot imunoprecipitated dimethyl Histone H3 (Lys36) in HeLa cell lysate.
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in HeLa acid extract.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected dimethyl Histone H3 (Lys36) in 10 µg HeLa acid extract and demonstrated a loss of signal in 0.5 µg of unmethylated recombinant Histone H3.

Target description

~17 kDa observed

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Broad-spectrum fungal resistance in sorghum is conferred through the complex regulation of an immune receptor gene embedded in a natural antisense transcript.
Lee, et al.
Plant Cell, 34, 1641-1665 (2022)
Sanghun Lee et al.
The Plant cell, 28(7), 1640-1661 (2016-06-30)
Posttranslational modification of histones modulates gene expression affecting diverse biological functions. We showed that the Arabidopsis thaliana histone methyl transferases SET DOMAIN GROUP8 (SDG8) and SDG25 regulate pep1-, flg22-, and effector-triggered immunity as well as systemic acquired resistance. Genome-wide basal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service