Skip to Content
Merck
All Photos(1)

Key Documents

07-218

Sigma-Aldrich

Anti-Notch1 Antibody, extracellular domain

Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, mouse

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Gene Information

human ... NOTCH1(4851)

General description

The Notch protein family is a group of highly conserved proteins important in the determination of cell fate and maintenance of progenitors in many developmental systems. This family of proteins function both as membrane cell receptors and as transcription factors. Activation of Notch by cell-cell interactions causes a transcription inhibitory effect that enables inhibition of differentiation in some cells but not in others. As a consequence, some cells adopt a particular fate while other progenitors remain uncommitted. The Notch protein is important in cell fate during myogenesis, neurogenesis, oogenesis, and wing and eye development in Drosophila. In mammals, four Notch genes were identified (Notch 1-4) that are expressed in a wide variety of cells and play a crucial role in differentiation and development.

Specificity

Notch1

Immunogen

N-terminal GST fusion protein corresponding to residues 381-853 of rat Notch1

Application

Anti-Notch1 Antibody, extracellular domain is a high quality Rabbit Polyclonal Antibody for the detection of Notch1 & has been validated in WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
Used in mouse in the following publication: Gulino, R., et al, European Journal of Neuroscience, Vol. 31, pp. 1423–1434, 2010.

Quality

Routinely evaluated by immunoblot on RIPA lysates from L/N17 rat Notch1 transfected fibroblast cells

Target description

260kDa

Linkage

Replaces: 04-1046

Physical form

Format: Purified
Protein A chromatography
Protein A purified immunoglobulin presented in 0.1M Tris-Glycine, 0.15M NaCl, and 0.05% Sodium Azide, pH 7.4

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
L/N17 transfected Notch1 cell lysate

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Notch signaling: direct or what?
Weinmaster, G
Current Opinion in Genetics & Development, 8, 436-442 (1998)
C M Blaumueller et al.
Cell, 90(2), 281-291 (1997-07-25)
Previous models for signal transduction via the Notch pathway have depicted the full-length Notch receptor expressed at the cell surface. We present evidence demonstrating that the Notch receptor on the plasma membrane is cleaved. This cleavage is an evolutionarily conserved
NF-kappaB2 is a putative target gene of activated Notch-1 via RBP-Jkappa
Oswald, F, et al
Molecular and cellular biology, 18, 2077-2088 (1998)
G Struhl et al.
Cell, 93(4), 649-660 (1998-05-30)
The Drosophila Notch (N) gene encodes a conserved single-pass transmembrane receptor that transduces extracellular signals controlling cell fate. Here, we present evidence that the intracellular domain of Notch gains access to the nucleus in response to ligand, possibly through a
Bo Yan et al.
The Journal of biological chemistry, 277(33), 29760-29764 (2002-06-18)
Acid alpha-glucosidase (GAA) is a lysosomal enzyme that degrades glycogen. A deficiency of GAA is responsible for a recessively inherited myopathy and cardiomyopathy, glycogenosis type II. Previously, we identified an intronic repressor element in the GAA gene and demonstrated that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service