Skip to Content
Merck
All Photos(2)

Key Documents

07-214

Sigma-Aldrich

Anti-dimethyl-Histone H3 (Arg17) Antibody

Upstate®, from rabbit

Synonym(s):

H3R17me2, Histone H3 (di methyl R17), H3 histone family, member T, histone 3, H3, histone cluster 3, H3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, vertebrates

manufacturer/tradename

Upstate®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
inhibition assay: suitable (peptide)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

trimethylation (Arg17)

Gene Information

human ... H3C1(8350)

General description

Histone H3.1t (UniProt: Q16695; also known as H3/t, H3tm, H3/g) is encoded by the HIST3H3 (also known as H3FT) gene (Gene ID: 8290) in human. Histone H3 has two main variants, H3.1 and H3.3, which show different genomic localization patterns in animals. The H3.1 and H3.3 complexes also possess distinct histone chaperones, CAF-1 and HIRA, which play important role in mediating DNA-synthesis-dependent and -independent nucleosome assembly. It has been reported that Histone H3.1 serves as the canonical histone, which is incorporated during DNA replication, whereas H3.3 acts as the replacement histone that can be incorporated outside of S-phase during chromatin-disrupting processes like transcription. Histone H 3.1 is a core component of nucleosome that is present only in mammals and is usually enriched in acetylation of Lysine 15 and demethylation of lysine 10 (HeK9Me2). It is expressed during S phase, then expression decreases significantly as cell division slows down during the process of differentiation. Histone H 3.1 expression is shown to be replication dependent. It s presence at the site of UV-induced DNA damage has also been reported. It has also been shown that H3.1/H4 tetramers do not split and remain intact during replication dependent deposition of H3.1 variant. Dimethylation of H3 on Arginine 17 has been linked to gene activation and activation of this modification is linked to cell fate regulation and increases the pluripotency potential of stem cells. In pancreatic beta cells expression of Protein arginine methyltransferase 4 (PRMT4) and demethylation of Arginine 17 is shown to increase under high glucose conditions. (Ref.: Kim, JK et al. (2015). J. Mol. Endocrinol. 54(3); 315-24).

Specificity

Broad cross-reactivity expected based on sequence homology among species.
This rabbit polyclonal antibody specifically detects human Histone H3 dimethylated on Arginine 17.

Immunogen

BSA-conjugated linear peptide corresponding to 11 amino acids from the N-terminal region of human Histone H3 dimethylated on Arginine 17 (methionine removed).

Application

Anti-dimethyl-Histone H3 (Arg17) Antibody is a Rabbit Polyclonal Antibody for detection of dimethyl-Histone H3 (Arg17) also known as H3R17me2, Histone H3 (di methyl R17) & has been validated in ELISA, ICC peptide inhibition & WB.
Peptide Inhibition Assay Analysis/Western Blotting: A 1:7,500 dilution from a representative lot detected dimethyl-Histone H3 (Arg17) in Histone H3 with CARM1. The dimethylation was inhibited in the presence of peptide containing Di-methyl Arg17.
ELISA:
A previous lot of this antibody was used in ELISA. At dilutions greater than 1:2000 this antibody is specific for peptides containing dimethylated Arg17. No cross-reactivity detected with a peptide containing dimethylated Arg26.

Immunocytochemistry:
An independent laboratory showed positive immunostaining for CARM-1 specific methylation of Histone H3 in 3134 cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

Evaluated by Western Blotting in Histone H3 with CARM1.

Western Blotting Analysis: A 1:7,500 dilution of this antibody detected dimethyl-Histone H3 (Arg17) in Histone H3 treated with Histone-arginine methyltransferase (CARM1).

Target description

~17 kDa observed; 15.51 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Immunoaffinity Chromatography
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Calf thymus histone preparation, Acid HeLa cell extract.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Coactivator-associated arginine methyltransferase-1 enhances nuclear factor-kappaB-mediated gene transcription through methylation of histone H3 at arginine 17.
Miao, F; Li, S; Chavez, V; Lanting, L; Natarajan, R
Molecular Endocrinology null
Caroline S Dacwag et al.
Molecular and cellular biology, 29(7), 1909-1921 (2009-02-04)
Temporal regulation of gene expression is a hallmark of cellular differentiation pathways, yet the mechanisms controlling the timing of expression for different classes of differentiation-specific genes are not well understood. We previously demonstrated that the class II arginine methyltransferase Prmt5
Donghang Cheng et al.
Molecular cell, 25(1), 71-83 (2007-01-16)
The coactivator-associated arginine methyltransferase CARM1 is recruited by many different transcription factors as a positive regulator. To understand the mechanism by which CARM1 functions, we sought to isolate its substrates. We developed a small-pool screening approach for this purpose and
HuD interacts with survival motor neuron protein and can rescue spinal muscular atrophy-like neuronal defects.
Hubers, L; Valderrama-Carvajal, H; Laframboise, J; Timbers, J; Sanchez, G; Cote, J
Human Molecular Genetics null
Cell-type selective chromatin remodeling defines the active subset of FOXA1-bound enhancers.
Eeckhoute, J; Lupien, M; Meyer, CA; Verzi, MP; Shivdasani, RA; Liu, XS; Brown, M
Genome Research null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service