Skip to Content
Merck
All Photos(2)

Key Documents

07-1562

Sigma-Aldrich

Anti-EZH2 Antibody

serum, Upstate®

Synonym(s):

SETDB1, EC 2.1.1.43

SET domain bifurcated 1

ERG-associated protein with SET domain

ESET

Histone H3-K9 methyltransferase 4

H3-K9-HMTase 4

Lysine N-methyltransferase 1E

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, mouse, human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

GenBank accession no.

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... EZH2(2146)

General description

Polycomb protein group complexes are responsible for the regulation of hundreds of genes in mammals and insects. These proteins are responsible for the assembly and packaging of chromatin and for the induction of methylation on Histone H3. EZH2 (enhancer of zeste homologue 2), also known as Enx1, is a member of the polycomb group of proteins and is involved in cell cycle regulation. EZH2 represses transcription via trimethylation of histone H3 on Lys27 (H3K27) as indicated by the fact that RNAi-mediated knockdown of EZH2 resulted in a loss of H3K27 trimethylation. The expression of this protein in human tumors is aggressive for hormone refractory prostate cancer. In breast cancer EZH2 expression was increased in malignant tumors and promoted anchorage independent and invasive growth in vitro.. EZH2, also known as ENX-1, is ubiquitously expressed during early embryo genesis, and becomes restricted to the central and peripheral nervous systems and sites of fetal hematopoiesis during later development. In the adult, EZH2 is restricted to the spleen, testis and placenta. EZH2 and BMI-1 genes are co expressed in Reed-Sternberg cells of Hodgkin’s disease. Coexpression of BMI-1 and EZH2 is also associated with cycling cells and degree of malignancy in B-cell non-Hogkin’s lymphoma. EZH2 is involved in the progression of prostate cancer, and has been identified as a marker of aggressive breast cancer. Two transcript variants encoding distinct isoforms have been identified for this gene.

Specificity

Recognizes EZH2, Mr ~100 kDa

Immunogen

KLH-conjugated, His-tagged fusion protein corresponding to residues 406-746 of human EHZ2 (similar to enhancer of zeste (Drosophila) homolog 2). Based on accession # BC010858.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Histones
Use Anti-EZH2 Antibody (Rabbit Polyclonal Antibody) validated in WB to detect EZH2 also known as SET domain bifurcated 1, ERG-associated protein with SET domain.

Quality

Routinely evaluated by immunoblot analysis

Target description

95-100kDa

Linkage

Replaces: 04-1047

Physical form

Serum

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
A431 cell lysate

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alok Jha et al.
Cells, 9(4) (2020-04-23)
Apolipoprotein L1 (APOL1)-miR193a axis has been reported to play a role in the maintenance of podocyte homeostasis. In the present study, we analyzed transcription factors relevant to miR193a in human podocytes and their effects on podocytes' molecular phenotype. The motif

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service