Skip to Content
Merck
All Photos(2)

Key Documents

07-1487

Sigma-Aldrich

Anti-JAK3 Antibody

serum, from rabbit

Synonym(s):

Janus Kinase 3, Janus kinase 3 (a protein tyrosine kinase, leukocyte), Leukocyte janus kinase, Tyrosine-protein kinase JAK3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$524.00

$524.00


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$524.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$524.00


Estimated to ship on19 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... JAK3(3718)
mouse ... Jak3(16453)

General description

JAKs are non-receptor tyrosine kinases which couple to cytokine receptors, and are activated by cytokines. JAK kinases (JAK1, JAK2, JAK3, Tyk2) have a kinase domain and a second, pseudokinase domain which is not enzymatically active. The principal effectors of JAKs are the STATs, which are direct activators of transcription, but JAKs also signal to the Ras-MAP Kinase pathway, and through Tec, to the PI3 Kinase pathway. JAK3 is required for signaling through cytokine receptors which share a common γ chain. JAK2 is required for signaling through a different subset of cytokine receptors, while JAK1 is essential for signaling from the IL-2, IL-6 and interferon receptors. The stress-activated protein kinase 1 (SAPK) family is also referred to as the jun N-terminal kinase family in light of the substrate preference of these serine/threonine kinases. In addition to regulating many biological processes including proliferation and apoptosis, the JNK/SAPK family has been implicated in many neurodegenerative diseases including Alzheimer disease, Parkinson′s disease and Amyotrophic lateral sclerosis.

Specificity

Recognizes JAK3 (Janus Kinase 3).

Immunogen

Synthetic peptide corresponding to amino acids 782-797 of mouse JAK3 (SDPTPGIPSPRDELC).

Application

Detect JAK3 using this Anti-JAK3 Antibody validated for use in WB & IP.
Immunoprecipitation: 5-10 uL of a previous lot of this antibody was used in immunoprecipitation.

Optimal working dilutions must be determined by end user.
Research Category
Signaling
Research Sub Category
Immunological Signaling

Quality

Routinely evaluated by Western Blot on HeLa lysates.

Western Blot Analysis:
1:500 dilution of this lot detected JAK3 on 10 μg of HeLa lysates.

Target description

125 kDa

Linkage

Replaces: AB3806

Physical form

Rabbit polyclonal IgG whole antiserum in buffer containing sodium azide.

Storage and Stability

Stable for 1 year at -20°C in undiluted aliquots from date of receipt.
Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
HeLa cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lanhai Lü et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(7), 9120-9140 (2020-05-21)
Homeostasis between pro- and anti- inflammatory responses induced by bacteria is critical for the maintenance of health. In the oral cavity, pro-inflammatory mechanisms induced by pathogenic bacteria are well-established; however, the anti-inflammatory responses that act to restrain innate responses remain

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service