Skip to Content
Merck
All Photos(1)

Documents

07-1386

Sigma-Aldrich

Anti-Connective Tissue Growth Factor Antibody

from goat

Synonym(s):

Hypertrophic chondrocyte-specific protein 24, connective tissue growth factor, insulin-like growth factor-binding protein 8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

goat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CTGF(1490)

General description

Connective Tissue Growth Factor (CTGF) is a member of the CNN family of growth factors that are characterized as having cysteine-rich peptides with four specific domains. CTGF was originally identified in endothelial cells as a significant chemotactic and mitogenic factor. Research has revealed CTGF to have several biological properties which include chemotaxis, cell proliferation, promotion of adhesion and the formation of extracellular matrix. It is also necessary for such biological processes as wound healing, scar formation, fibrosis, embryogenesis and angiogenesis. In its role as an inducer of angiogenesis, CTGF has been associated with vascular endothelial growth factor (VEGF). As a result of its pro-fibrotic properties, CTGF is thought to be a promising target for therapy in conditions involving pathological fibrosis such as chronic fibrotic disease, chronic kidney disease and proliferative diabetic retinopathy (PDR). Cell types found to express CTGF are endothelial cells, fibroblasts and vascular smooth-muscle cells.

Specificity

Primate (98% sequence homolgy). Monkey (97% sequence homology). Porcine and Bovine (93% sequence homology). Rat and mouse (91% sequence homology).
This antibody recognizes the mature chain of Connective tissue growth factor.

Immunogen

Epitope: Mature chain
Recombinant protein corresponding to the mature chain of human Connective tissue growth factor.

Application

Anti-Connective Tissue Growth Factor Antibody detects level of Connective Tissue Growth Factor & has been published & validated for use in WB.
Research Category
Signaling
Research Sub Category
Growth Factors & Receptors

Quality

Evaluated by Western Blot in untreated and TGFb1 treated NRK cell lysates.

Western Blot Analysis: 5 µg/mL of this antibody detected Connective tissue growth factor on 10 µg of untreated and TGFb1 treated NRK cell lysates.

Target description

In Western Blot, full length CTGF is observed at 38 kDa and CTGF N&C domain is observed at 19 kDa

Physical form

Format: Purified
Protein G Purified
Purified goat polyclonal in buffer containing Tris-Glycine pH 8.0 with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Untreated and TGFb1 treated NRK cell lysates

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service