Skip to Content
Merck
All Photos(2)

Key Documents

07-1364

Sigma-Aldrich

Anti-ECT2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

epithelial cell transforming sequence 2 oncogene, Epithelial cell-transforming sequence 2 oncogene, protein ECT2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$632.00

$632.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$632.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$632.00


Estimated to ship on30 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

pig, human

species reactivity (predicted by homology)

porcine (based on 100% sequence homology), rabbit (based on 100% sequence homology), monkey (based on 100% sequence homology), opossum (based on 100% sequence homology), rat (based on 100% sequence homology), mouse (based on 100% sequence homology), bovine (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ECT2(1894)

General description

Epithelial cell-transforming sequence 2 oncogene (ECT2) is a guanine nucleotide exchange factor or GEF that belongs to the Dbl family of Rho GEFs. ECT2 is composed of 2 BRCT domains within the N-terminus, a DBL-homology domain, and a PH domain. Studies have shown ECT2 to localize to both the cell cortex and central spindles, and that it is a crucial part of cleavage furrow formation during cytokenises. Overexpression of cleaved ECT2, containing only the N-terminal domain, has been shown to cause multinucleation. As a GEF it has been shown to bind specifically to Rac proteins, RhoA, and RhoC.

Specificity

This antibody recognizes ECT2 at the
C-terminus.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to human ECT2 at the C-terminus.

Application

Anti-ECT2 Antibody is an antibody against ECT2 for use in WB & IC.
Research Category
Signaling
Research Sub Category
G-proteins
Western Blot (Snap i.d.) Analysis: 5 µg/mL from a previous lot detected ECT2 in A549 cell lysate.
Immunocytochemistry Analysis: A previous lot was used by an independent laboratory on Heya8 cells. (Der Channing, University of North Carolina at Chapel Hill, NC, [email protected])

Quality

Evaluated by Western Blot in A549 cell lysate.
Western Blot Analysis: 1 µg/mL of this antibody detected ECT2 on 10 µg of A549 cell lysate.

Target description

~ 105 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
A549 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ming Yi et al.
Experimental hematology & oncology, 11(1), 107-107 (2022-12-27)
Breast cancer is the most common malignancy and the second leading cause of cancer-related death in women. Recent studies have indicated that aberrant activation of Rho GTPases relates to the malignant properties of breast cancer cells. As the guanine nucleotide
Laurent Guillemot et al.
Molecular biology of the cell, 25(13), 1995-2005 (2014-05-09)
The regulation of Rho-family GTPases is crucial to direct the formation of cell-cell junctions and tissue barriers. Cingulin (CGN) and paracingulin (CGNL1) control RhoA activation in epithelial cells by interacting with RhoA guanidine exchange factors. CGNL1 depletion also inhibits Rac1
Bin Liu et al.
Translational oncology, 11(3), 700-714 (2018-04-10)
Pancreatic cancer is the fourth leading cause for cancer-related death, and early diagnosis is one key to improve the survival rate of this disease. Molecular biomarkers are an important method for diagnostic use in pancreatic cancer. We used data from
Xianguang Bai et al.
Cancer management and research, 10, 4039-4050 (2018-10-16)
Epithelial cell transforming sequence 2 (ECT2) is a guanine nucleotide exchange factor, which is involved in cell division regulation and cell cycle modulation. Recent evidence indicates that ECT2 is overexpressed in many human cancers. However, the exact prognostic value of
Danielle R Cook et al.
Genes & cancer, 2(10), 932-942 (2012-06-16)
Ect2 is a member of the human Dbl family of guanine nucleotide exchange factors (RhoGEFs) that serve as activators of Rho family small GTPases. Although Ect2 is one of at least 25 RhoGEFs that can activate the RhoA small GTPase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service