Skip to Content
Merck
All Photos(2)

Key Documents

07-1221

Sigma-Aldrich

Anti-PP2A-B′ (B56) Antibody

from rabbit

Synonym(s):

PP2A phosphatase activator, PP2A, subunit B′, PP2A, subunit B′, PR53 isoform, Phosphotyrosyl phosphatase activator, protein phosphatase 2A activator, regulatory subunit 4, protein phosphatase 2A, regulatory subunit B′, protein phosphatase 2A, regulatory

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$572.00

$572.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$572.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$572.00


Estimated to ship on26 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, human, mouse, bovine

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PTPA(5524)

General description

PP2A is a heterotrimeric type 2 Ser/Thr phosphatase, composed of a structural subunit (A), a regulatory subunit (B), and a catalytic subunit (C). The structural A and catalytic C subunits form the constitutive core of the enzyme, which associates with one of a large number of regulatory B subunits. There are multiple isotypes (encoded by distinct genes) of the B subunit, grouped into three families: B/PR55, B′/PR61, and B′′/PR72 . The prototypical B (or PR55) family of subunits is encoded by three genes, Balpha, Bbeta and Bgamma . The B′ (or B56) family consists of at least seven splice variants derived from five genes. RNA′s for the two members of the B′′ family, PR72 and PR130, arise by alternative splicing or use of alternate transcription start sites from the same gene. Primary structures as well as functions of B and B′ subunits are conserved from yeast to mammals. PTPA is a member of the B′ family that is required for activation of PP2A by an undefined mechanism. PP2A is distinguishable from PP2B/Calcineurin and PP2C by its partial activity in the absence of divalent cations, and its high sensitivity to Okadaic Acid. It is also thought to be the principal downmodulator of Ser/Thr kinase cascades.

Specificity

This antibody is specific to PP2A-B’ subunits.

Immunogen

Epitope: Internal
The immunogen used for the purified peptide conjugated to KLH corresponds to the sequence NH2-Val-Ser-Ser-Pro-His-Phe-Gln-Val-Ala-Glu- Arg-Ala-Leu-Tyr-COOH.

Application

Detect PP2A-B’ (B56) using this Anti-PP2A-B’ (B56) Antibody validated for use in WB.
Research Category
Signaling
Research Sub Category
PI3K, Akt, & mTOR Signaling

Kinases & Phosphatases

Quality

Routinely evaluated by Western Blot on HeLa lysates.

Western Blot Analysis: 1:500-1:1,500 dilution of this antibody detected PP2A-B′ (B56) in 10 μg of multiple cell lysates.

Target description

~56 kDa

Linkage

Replaces: AB4088

Physical form

Format: Purified
Purified
Purified rabbit polyclonal IgG in PBS with 0.08% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 1 year from date of receipt.
Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
HeLa Cell Lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li Fan et al.
Oncotarget, 7(39), 63887-63900 (2016-09-03)
The regulation of the ubiquitously expressed protein phosphatase 2A (PP2A) is essential for various cellular functions such as cell proliferation, transformation, and fate determination. In this study, we demonstrate that the highly conserved protein in mammals, designated FAM122A, directly interacts

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service