Skip to Content
Merck
All Photos(2)

Key Documents

06-559

Sigma-Aldrich

Anti-Pyk2 Antibody

Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... PTK2B(2185)

Specificity

Recognizes Pyk2, and its degradation products; does not cross-react with FAK.

Immunogen

GST-fusion protein containing amino acids 726-863 of human Pyk2

Application

Research Category
Signaling
Research Sub Category
Cytoskeletal Signaling
This Anti-Pyk2 Antibody is validated for use in IC, IP, WB for the detection of Pyk2.

Quality

routinely evaluated by immunoblot on mouse brain cytosolic RIPA lysates

Target description

125 kDa

Physical form

Format: Purified
Protein A Purified immunoglobulin in 0.07M Tris-glycine, pH 7.4, 0.105 M NaCl, 0.035% sodium azide as a preservative.
Protein A purified

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse brain cytosolic RIPA lysates

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydroxamic acid derivatives of mycophenolic acid inhibit histone deacetylase at the cellular level.
Daniela I Batovska,Dong Hoon Kim,Shinya Mitsuhashi,Yoon Sun Cho,Ho Jeong Kwon,Makoto Ubukata
Bioscience, Biotechnology, and Biochemistry null
Pyk2 inhibition of p53 as an adaptive and intrinsic mechanism facilitating cell proliferation and survival.
Ssang-Taek Lim,Nichol L G Miller,Ju-Ock Nam,Xiao Lei Chen,Yangmi Lim,David D Schlaepfer
The Journal of Biological Chemistry null
Shinichi Nakano et al.
Scientific reports, 9(1), 4236-4236 (2019-03-14)
Excessive osteoclastic bone erosion disrupts normal bone remodeling and leads to bone loss in many skeletal diseases, including inflammatory arthritis, such as rheumatoid arthritis (RA) and psoriatic arthritis, periodontitis and peri-prosthetic loosening. Functional control of osteoclasts is critical for the
Mad3 negatively regulates B cell differentiation in the spleen by inducing Id2 expression.
Gore, Y; Lantner, F; Hart, G; Shachar, I
Molecular Biology of the Cell null
Maja Beus et al.
ChemistryOpen, 7(8), 624-638 (2018-08-29)
We report the synthesis of SAHAquines and related primaquine (PQ) derivatives. SAHAquines are novel hybrid compounds that combine moieties of suberoylanilide hydroxamic acid (SAHA), an anticancer agent with weak antiplasmodial activity, and PQ, an antimalarial drug with low antiproliferative activity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service