Skip to Content
Merck
All Photos(1)

Key Documents

06-495

Sigma-Aldrich

Anti-PDGFRβ Antibody

Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse, monkey

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PDGFRB(5159)

Specificity

Recognizes PDGF type B receptor.

Immunogen

peptide (PAPRAEAEDSFL) corresponding to the amino acids 1095-1106 of human PDGF type B receptor

Application

Anti-PDGFRβ Antibody is an antibody against PDGFRβ for use in IP & WB.
Research Category
Signaling
Research Sub Category
Growth Factors & Receptors

Quality

routinely evaluated by immunoblot on mouse 3T3/A31 cell RIPA lysate

Target description

170/190 kDa

Linkage

Replaces: 04-397

Physical form

Format: Purified
Protein A Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105 M NaCl, 0.035% sodium azide as a preservative.
Protein G Purified

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Positive Antigen Control: Catalog #12-305, 3T3/A31 lysate. Add 2.5 μL of 2-mercapto-ethanol/100 μL of lysate and boil for 5 minutes to reduce the preparation. Load 20 μg of reduced lysate per lane for minigels.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R L Panek et al.
Arteriosclerosis, thrombosis, and vascular biology, 17(7), 1283-1288 (1997-07-01)
Platelet-derived growth factor (PDGF) receptor gene expression has previously been demonstrated in balloon-injured rat carotid arteries to be regulated during repair of carotid injury. In the present study we showed that PDGF receptor protein expression and phosphorylation are changed over
R M Kypta et al.
Cell, 62(3), 481-492 (1990-08-10)
We have examined the interaction between the platelet-derived growth factor (PDGF) receptor and three src family tyrosine kinases, pp60c-src, p59fyn, and pp62c-yes. The kinase activities of all three enzymes were elevated after PDGF stimulation of quiescent fibroblasts, coincident with association
Platelet-derived growth factor-stimulated secretion of basement membrane proteins by skeletal muscle occurs by tyrosine kinase-dependent and -independent pathways.
Albrecht, D E and Tidball, J G
The Journal of Biological Chemistry, 272, 2236-2244 (1997)
Paul W Manley et al.
Biochimica et biophysica acta, 1804(3), 445-453 (2009-11-20)
As a drug used to treat imatinib-resistant and -intolerant, chronic and advanced phase chronic myelogenous leukaemia, nilotinib is well characterised as a potent inhibitor of the Abl tyrosine kinase activity of wild-type and imatinib-resistant mutant forms of BCR-Abl. Here we
Tammara L Watts et al.
Journal of translational medicine, 14(1), 337-337 (2016-12-10)
The robust desmoplasia associated with head and neck squamous cell carcinoma (HNSCC) suggests that the tumor microenvironment may be an important component in the pathophysiology of this cancer. Moreover, the high recurrence rate and poor clinical response to chemotherapy and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service