Skip to Content
Merck
All Photos(3)

Key Documents

06-1082-I

Sigma-Aldrich

Anti-IL-27A Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Interleukin-27 subunit alpha, IL-27 subunit alpha, IL-27-A, IL27-A, Interleukin-30, p28, IL-27A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$520.00

$520.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$520.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$520.00


Estimated to ship on20 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... IL27(246778)

General description

Interleukin 27 (IL-27) is a heterodimeric cytokine belonging to the IL-12 family that is composed of two subunits: Epstein-Barr virus (EBV)-induced gene 3 (EBI3) (also known as IL-27B) and IL27-p28 (known as IL-27A). IL-27 has pro- and anti-inflammatory properties, that can regulate T helper cell development, suppress T-cell proliferation, stimulate cytotoxic T cell activity, induce isotype switching in B-cells, and that has diverse effects on innate immune cells. IL-27 is secreted by antigen-presenting cells and plays an important function in regulating the activity of B and T lymphocytes. IL-27 suppresses the expression of GATA3, the inhibitor of TH1 cell development. In addition to suppressing cytokine production, IL-27 also antagonizes the effects of some cytokines such as IL6 through direct effects on T cells. Another important role of IL-27 is its anti-tumor activity as well as its anti-angiogenic activity with activation of production of anti-angiogenic chemokines such as IP-10/CXCL10 and MIG/CXCL9. In vein endothelial cells, it induces IRF1/interferon regulatory factor 1 and increases the expression of MHC class II transactivator/CIITA with resulting up-regulation of major histocompatibility complex class II. IL-27 also demonstrates antiviral activity with inhibitory properties on HIV-1 replication.

Immunogen

KLH-conjugated linear peptide corresponding to human IL-27A.

Application

Anti-IL-27A Antibody is an antibody against IL-27A for use in Western Blotting, Immunohistochemistry and Flow Cytometry.
Immunohistochemistry Analysis: A 1:50-1,000 dilution from a representative lot detected IL-27A in human placenta and human tonsil tissue.
Flow Cytometry Analysis: 0.5 µg of this antibody from a representative lot detected IL-27A in 1X10E6 staph enterotoxin B treated human peripheral blood mononuclear cells.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

Evaluated by Western Blotting in human placenta tissue lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected IL-27A in 10 µg of human placenta tissue lysate.

Target description

~27 kDa observed. Uncharacterized band(s) may appear in some lysates.

Linkage

Replaces: 06-1082

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service