Skip to Content
Merck
All Photos(1)

Key Documents

06-018

Sigma-Aldrich

Anti-ROR2 Antibody, CT

from rabbit

Synonym(s):

Neurotrophic tyrosine kinase, receptor-related 2, neurotrophic tyrosine kinase receptor-related 2, receptor tyrosine kinase-like orphan receptor 2, tyrosine-protein kinase transmembrane receptor ROR2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ROR2(4920)

General description

Receptor tyrosine kinase-like orphan receptor 2, also known as ROR2, is a member of the Tyr protein kinase family and ROR subfamily of cell surface receptors. Located in the transmembrane, ROR2 is highly expressed during early embryonic development and is associated with chondrocyte formation, cartilage and growth plate maturity. Expression levels of ROR2 subside after 16 days and occur minimally in adults. ROR2 is post translationaly modified by ATM or ATR phosphorylation when DNA damage occurs. Mutations in ROR2 result in brachydactyly type B1 (BDB1), characterized by either hypoplasia or aplasia of the distal phalanges and nails. ROR2 is also associated with recessive Robinow syndrome (RRS), resulting in shortening of limbs, facial dysmorphia and spinal defects.

Specificity

This antibody recognizes the C-terminal region of ROR2.

Immunogen

Epitope: C-terminus
KLH-conjugated synthetic linear peptide corresponding to the C-terminal region of ROR2

Application

Anti-ROR2 Antibody, C-terminus detects level of ROR2 & has been published & validated for use in IH(P).
Research Category
Inflammation & Immunology
Research Sub Category
Immunological Signaling

Quality

Evaluated by immunohistochemistry (paraffin) in human gastric mucosa.

Immunohistochemistry(paraffin): 1:50-1:200 dilution of this antibody detected ROR2 in human gastric mucosa.

Target description

~105 kDa

Physical form

Format: Purified
Protein G Purified
Purified rabbit polyclonal in buffer containing PBS and 0.09% sodium azide.

Storage and Stability

Stable for 1 year at 2-8ºC from date of receipt.

Analysis Note

Control
Human gastric mucosa

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dominika Maciejewska-Markiewicz et al.
Nutrients, 14(20) (2022-10-28)
Non-alcoholic fatty liver disease (NAFLD) is a wide spectrum condition characterized by excessive liver fat accumulation in people who do not abuse alcohol. There is no effective medical treatment for NAFLD; therefore, most important recommendations to reduce liver steatosis are
Xiaonan Liu et al.
Nature protocols, 15(10), 3182-3211 (2020-08-12)
Affinity purification coupled with mass spectrometry (AP-MS) and proximity-dependent biotinylation identification (BioID) methods have made substantial contributions to interaction proteomics studies. Whereas AP-MS results in the identification of proteins that are in a stable complex, BioID labels and identifies proteins
Izabela N Faria Gomes et al.
Molecules (Basel, Switzerland), 24(21) (2019-11-07)
Plant-based compounds are an option to explore and perhaps overcome the limitations of current antitumor treatments. Annona coriacea Mart. is a plant with a broad spectrum of biological activities, but its antitumor activity is still unclear. The purpose of our

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service